Skip to Content
Merck
All Photos(1)

Key Documents

361275

Sigma-Aldrich

3′-Deoxy-3′-fluorothymidine

97%

Synonym(s):

3′-Fluoro-2′,3′-dideoxythymidine, 3′-Fluorodeoxythymidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13FN2O4
CAS Number:
Molecular Weight:
244.22
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

176-178 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1=CN([C@H]2C[C@H](F)[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H13FN2O4/c1-5-3-13(10(16)12-9(5)15)8-2-6(11)7(4-14)17-8/h3,6-8,14H,2,4H2,1H3,(H,12,15,16)/t6-,7+,8+/m0/s1

InChI key

UXCAQJAQSWSNPQ-XLPZGREQSA-N

Gene Information

human ... TK2(7084)

Looking for similar products? Visit Product Comparison Guide

Application

Promising antiviral agent possessing activity similar to other 3′-deoxy-3′-substituted thymidines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael S Enslow et al.
Clinical nuclear medicine, 37(9), 854-861 (2012-08-15)
The objective was to compare F-fluorodeoxyglucose (FDG) and F-fluorothymidine (FLT) PET in differentiating radiation necrosis from recurrent glioma. Visual and quantitative analyses were derived from static FDG PET and static and dynamic FLT PET in 15 patients with suspected recurrence
Albert J S Idema et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 53(12), 1904-1910 (2012-10-19)
3'-deoxy-3'-(18)F-fluorothymidine ((18)F-FLT) is a radiopharmaceutical depicting tumor cell proliferation with PET. In malignancies of the lung, breast, head and neck, digestive tract, brain, and other organs, quantitative assessment of (18)F-FLT targeting has been shown to correlate with the proliferation marker
Bianca A W Hoeben et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(4), 532-540 (2013-01-25)
This prospective study used sequential PET with the proliferation tracer 3'-deoxy-3'-(18)F-fluorothymidine ((18)F-FLT) to monitor the early response to treatment of head and neck cancer and evaluated the association between PET parameters and clinical outcome. Forty-eight patients with head and neck
Camilo Garcia et al.
Current opinion in oncology, 24(6), 748-755 (2012-08-24)
(18)F-fluoro-2-deoxy-D-glucose PET-computed tomography (CT) has a major clinical impact in oncology not only for diagnostics but also for staging, response assessment, and relapse detection. However, several other PET tracers are needed for a more complete molecular characterization of cancers which
Matthias Scheffler et al.
PloS one, 8(1), e53081-e53081 (2013-01-12)
3'-deoxy-3'-[(18)F]fluoro-L-thymidine (FLT) and 2'-deoxy-2'-[(18)F]fluoro-D-glucose (FDG) are used to visualize proliferative and metabolic activity of tumors. In this study we aimed at evaluating the prognostic value of FLT and FDG uptake measured by positron emission tomography (PET) in patients with metastatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service