Skip to Content
Merck
All Photos(3)

Documents

327050

Sigma-Aldrich

Silver

shot, 1-3 mm, ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
EC Number:
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

shot

resistivity

1.59 μΩ-cm, 20°C

particle size

1-3 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Application

Silver (Ag) based shot can be used for a variety of applications such as:
  • synthesis of silver nanoparticles
  • formation of an alloy with tin, silver, and copper for lead free soldering
  • preparation of metallic colloids
  • fabrication of metalorganic semiconducting films for surface-enhanced Raman scattering (SERS) application

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Metal?Organic Semiconductor Nanostructures for Surface-Enhanced Raman Scattering
Alessio P, et al.
Applied Spectroscopy, 65(2), 152-158 (2011)
Simultaneous preparation of SERS-active metal colloids and plates by laser ablation
Lee I, et al.
Journal of Raman Spectroscopy, 32(11), 947-952 (2001)
Tin-silver-copper eutectic temperature and composition
Loomans ME and Fine ME
Metallurgical and Materials Transactions A: Physical Metallurgy and Materials Science, 31(4), 1155-1162 (2000)
Synthesis of spherical silver nanoparticles by digestive ripening, stabilization with various agents, and their 3-D and 2-D superlattice formation
Smetana AB, et al.
Journal of Colloid and Interface Science, 284(2), 521-526 (2005)
R Nirmala et al.
Journal of nanoscience and nanotechnology, 13(7), 4686-4693 (2013-08-02)
We report on the preparation and characterization of polyurethane (PU) nanofibers containing silver (Ag) nanoparticles were synthesized by using electrospinning. Two different approaches were adopted to incorporate the Ag nanoparticles in to PU nanofibers. In the first approach, a homogeneous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service