Skip to Content
Merck
All Photos(1)

Key Documents

QC1132

Supelco

Trace Metals 1-WP

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 20 mL

manufacturer/tradename

RTC QCI-034-1

concentration

in 5% HNO3

technique(s)

AAS: suitable
ICP: suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Intended as a Quality Control Check sample for, but not limited to validating calibration curves, method validation, analyst training, and matrix spikes.

This sample is not intended for calibration purposes.
Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Arsenic
    Beryllium
    Cadmium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Lithium
    Manganese
    Mercury
    Nickel
    Selenium
    Vanadium
    Zinc
    Aluminum
    See All (16)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service