All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H9N5O4S
CAS Number:
Molecular Weight:
223.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
>300 °C (lit.)
SMILES string
OS(O)(=O)=O.Nc1ncnc(N)c1N
InChI
1S/C4H7N5.H2O4S/c5-2-3(6)8-1-9-4(2)7;1-5(2,3)4/h1H,5H2,(H4,6,7,8,9);(H2,1,2,3,4)
InChI key
RKJICTKHLYLPLY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
4,5,6-Triaminopyrimidine sulfate was converted to 4,5,6-triaminopyrimidine sulfite by formylation with C13 labeled formic acid that was used in the preparation of adenine labeled with C13.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Exchange between a Formamido Group and Formamide, Studied with C13, 1.
Journal of the American Chemical Society, 71(6), 2246-2247 (1949)
J J Gato-Peciña et al.
Revista espanola de fisiologia, 46(4), 343-352 (1990-12-01)
Passive absorption of D-Galactose (in the presence of 0.5 mM phlorizin), 2-deoxy-D-glucose and D-Mannitol by rat jejunum has been measured in vivo by perfusion of an intestinal segment with recirculation, along successive absorption periods of 5 or 10 min duration.
N E Rusakova et al.
Gigiena truda i professional'nye zabolevaniia, (9)(9), 43-45 (1989-01-01)
Photometric procedures of monitoring the content of adenine synthesis intermediate products were developed and optimum conditions of air sample analysis and selection determined. Thus it became possible to establish general exposure levels for (phenylazo) malondinitryl and 4,5,6 triaminopyrimidine sulfate, sensitivity
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service