Skip to Content
Merck
All Photos(2)

Key Documents

303232

Sigma-Aldrich

3-Nitro-2-pyridinesulfenyl chloride

95%

Synonym(s):

(3-Nitro-2-pyridyl)sulfenyl chloride, 2-(Chlorosulfanyl)-3-nitropyridine, 3-Nitropyridinyl-2-sulfenyl chloride, [(3-Nitropyridin-2-yl)sulfanyl]chlorane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3ClN2O2S
CAS Number:
Molecular Weight:
190.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

205 °C (dec.) (lit.)

solubility

dichloromethane: soluble(lit.)

functional group

nitro

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cccnc1SCl

InChI

1S/C5H3ClN2O2S/c6-11-5-4(8(9)10)2-1-3-7-5/h1-3H

InChI key

WTKQMHWYSBWUBE-UHFFFAOYSA-N

General description

The 3-nitro-2-pyridinesulphenyl (Npys) moiety is useful as a protecting-activating group for cysteine, particularly in the synthesis of cyclic and unsymmetrical disulfides. The stability of NpysCl was studied in various solvents.

Application

3-Nitro-2-pyridinesulfenyl chloride (NpysCl) was employed as the starting material for the synthesis of N-, O- and S-Npys-protected amino acid.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Matsueda et al.
Peptide research, 5(5), 262-264 (1992-09-01)
Two recent reports on the partial lability of the 3-nitro-2-pyridinesulfenyl (Npys) thiol protecting group towards 1-hydroxy-benzotriazole (HOBt) have prompted a rechecking of the chemical behavior of this group. Using both soluble and polymer-bound forms of Cys(Npys) as test materials, the
R G Simmonds et al.
International journal of peptide and protein research, 43(4), 363-366 (1994-04-01)
The 3-nitro-2-pyridinesulphenyl (Npys) moiety is finding increasing utility as a protecting-activating group for cysteine, particularly in the synthesis of cyclic and unsymmetrical disulfides using the Boc strategy. This chemistry has been extended to peptides assembled by the Fmoc strategy. N-Terminal
S Rajagopalan et al.
International journal of peptide and protein research, 45(2), 173-179 (1995-02-01)
TASPs (template-assembled synthetic peptides) are generated by the covalent attachment of linear peptides to a common peptide backbone, thus generating larger synthetic peptides/proteins with prefolded structure. In this work we present a strategy for the synthesis of a heterotemplate-assembled synthetic
O Rosen et al.
International journal of peptide and protein research, 35(6), 545-549 (1990-06-01)
The hydroxylic side-chain functional groups of serine, threonine, hydroxproline and tyrosine, the alpha and epsilon-amino moieties of lysine and the thiol group of cysteine were masked by the 3-nitro-2-pyridinesulfenyl (Npys) protecting group. Deprotection was mildly affected by thiolysis with either
Vanessa Barcelo-Bovea et al.
Cancers, 12(5) (2020-05-18)
The delivery of Cytochrome c (Cyt c) to the cytosol stimulates apoptosis in cells where its release from mitochondria and apoptotic induction is inhibited. We developed a drug delivery system consisting of Cyt c nanoparticles decorated with folate-poly(ethylene glycol)-poly(lactic-co-glycolic acid)-thiol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service