Skip to Content
Merck

D5767

2,2′-Dithiodipyridine

powder

Synonym(s):

2,2′-Dipyridyl disulfide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H8N2S2
CAS Number:
Molecular Weight:
220.31
UNSPSC Code:
12352112
NACRES:
NA.26
PubChem Substance ID:
EC Number:
218-343-1
Beilstein/REAXYS Number:
154629
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

mp

56-58 °C (lit.)

storage temp.

2-8°C

SMILES string

S(Sc1ccccn1)c2ccccn2

InChI

1S/C10H8N2S2/c1-3-7-11-9(5-1)13-14-10-6-2-4-8-12-10/h1-8H

InChI key

HAXFWIACAGNFHA-UHFFFAOYSA-N

Application

2,2′-Dithiodipyridine has been used in the reconstitution of the reduced interchain disulfide bonds of monolconal antibodies. It may be used as a medium component for T cell hybridoma cell culture and as thiol crosslinking agent.

Biochem/physiol Actions

2,2′-Dithiodipyridine mediates inter-unit disulfide cross-linking. It interacts with carbon nucleophiles to yield thiopyridyl derivatives.


Still not finding the right product?

Explore all of our products under 2,2′-Dithiodipyridine


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
D5767-5G04061832407739
D5767-1G04061833586792
D5767-25G04061833586808