288047
Triethyltin bromide
97%
Synonym(s):
Bromotriethylstannane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.526 (lit.)
bp
222-224 °C (lit.)
density
1.63 g/mL at 25 °C (lit.)
SMILES string
CC[Sn](Br)(CC)CC
InChI
1S/3C2H5.BrH.Sn/c3*1-2;;/h3*1H2,2H3;1H;/q;;;;+1/p-1
InChI key
KQPIFPBKXYBDGV-UHFFFAOYSA-M
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
210.2 °F - closed cup
Flash Point(C)
99 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Neurobehavioral toxicology and teratology, 3(3), 285-293 (1981-01-01)
On Day 5 of postnatal life, rat pups received a single injection of triethyltin and were later tested for a variety of developmental and adult behaviors. A within-litter dosing design was used with one male and one female from each
Toxicology and applied pharmacology, 82(1), 87-93 (1986-01-01)
A single injection of trimethyltin chloride (TMT; 2, 4, or 6 mg/kg) is shown to produce a frequency-specific, dose-dependent auditory impairment, as well as to decrease the amplitude of the acoustically elicited startle response, in exposed rats. This finding stands
On the role of carbonic anhydrase in the anticonvulsant effects of triethyltin (TET).
Experientia, 38(7), 824-826 (1982-07-15)
Interactions of triethyltin bromide with components of the red cell.
Toxicology and applied pharmacology, 58(1), 67-75 (1981-03-30)
Neurotoxicology, 5(4), 9-22 (1984-01-01)
Triethyltin (TET) has been shown to be neurotoxic when injected on postnatal day (PND) 5. In the present experiment we examined the toxicity of a single exposure to TET at several postnatal ages. Rat pups were injected ip with 0
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service