Skip to Content
Merck
All Photos(1)

Key Documents

146498

Sigma-Aldrich

Trimethyltin chloride

Synonym(s):

Chlorotrimethylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnCl
CAS Number:
Molecular Weight:
199.27
Beilstein:
3535111
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

crystals

mp

37-39 °C (lit.)

SMILES string

C[Sn](C)(C)Cl

InChI

1S/3CH3.ClH.Sn/h3*1H3;1H;/q;;;;+1/p-1

InChI key

KWTSZCJMWHGPOS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Trimethyltin chloride is an organotin reagent widely used in transferring trimethylstannyl groups onto the substrates to synthesize various organostannanes. Trimethylstannyl compounds derived from this reagent, are extensively used in the palladium-catalyzed Stille coupling reactions.

Application

Trimethyltin chloride can be used as a precursor to synthesize trimethyltin hydride, cyanide, methoxide, azide, and lithium compounds.
It can also be used as a reagent to prepare:
  • Organotrimethyltin derivatives by reacting with organocopper compounds via transmetalation reaction.
  • Acetophenone by palladium-catalyzed coupling reaction with benzoyl chloride.
  • Optically active propargyl trimethylstannane by treating with chiral allenyltitanium.
  • Trimethylstannyl nucleophiles, which are applicable in the formation of Sn-C bonds via SN2 reactions, SRN1 reactions, and halogen-metal exchanges.
  • Carbocycles by reacting with unactivated dienes or trienes via radical-mediated carbocyclization reaction in the presence of NaBH3CN and a catalytic amount of AIBN.

Me3SnCl can also be used as a Lewis acid catalyst in asymmetric allylic alkylation reactions.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Palladium-catalyzed coupling of tetraorganotin compounds with aryl and benzyl halides. Synthetic utility and mechanism.
Milstein D and Stille J K
Journal of the American Chemical Society, 101(17), 4992-4998 (1979)
Chlorotrimethylstannane.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2009)
Chlorotrimethylstannane
Yoshinori Yamamoto, et al.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2007)
Replacing alkoxy groups with alkylthienyl groups: a feasible approach to improve the properties of photovoltaic polymers.
Huo H, et al.
Angewandte Chemie (International Edition in English), 50(41), 9697-9702 (2011)
Yu Xi et al.
Journal of pineal research, 67(3), e12596-e12596 (2019-07-25)
Trimethyltin chloride (TMT) is a potent neurotoxin that causes neuroinflammation and neuronal cell death. Melatonin is a well-known anti-inflammatory agent with significant neuroprotective activity. Male C57BL/6J mice were intraperitoneally injected with a single dose of melatonin (10 mg/kg) before exposure to TMT (2.8 mg/kg, ip). Thereafter, the mice received

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service