Skip to Content
Merck
All Photos(2)

Documents

282782

Sigma-Aldrich

Platinum(II) acetylacetonate

97%

Synonym(s):

2,4-Pentanedione platinum(II) derivative, Pt(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(C5H7O2)2
CAS Number:
Molecular Weight:
393.29
Beilstein:
4136189
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

reaction suitability

core: platinum
reagent type: catalyst

mp

249-252 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Pt]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Pt/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

KLFRPGNCEJNEKU-FDGPNNRMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Platinum(II)acetylacetonate is widely used as a Ptsource and precursor in the preparation of nanocatalysts and films. It is alsoused as a catalyst in N-allylation reactions.

Application

Platinum(II)acetylacetonate can be used as an efficient catalyst:
  • In the preparation of N-allylanilines by the direct use of allylic alcohols. Anilines with electron-donating groups give good yields.
  • For selective N-allylation of 2,3-disubstituted indoles in water.
Pt-loaded ZnO thick films can be prepared by flame spray pyrolysis method using zinc naphthenate and platinum(II) acetylacetonate as precursors. These sensing films can be used for acetylene detection and show good selectivity towards carbon monoxide, hydrogen, and methane.

It can be used as a starting material in the preparation of platinum-lead nanoparticles supported on carbon. The PtPb/C catalyst exhibits enhanced electrochemical activity towards methanol and formic acid oxidation.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry Letters (Jpn), 33, 658-659 (2004)
Chemistry of Materials, 18, 6414-6414 (2006)
Xiaochen Shen et al.
Nature communications, 9(1), 4485-4485 (2018-10-28)
Understanding the growth pathway of faceted alloy nanoparticles at the atomic level is crucial to morphology control and property tuning. Yet, it remains a challenge due to complexity of the growth process and technical limits of modern characterization tools. We
Jin Wang et al.
Nanoscale research letters, 14(1), 11-11 (2019-01-10)
Electrochemical sensors have the high sensitivity, fast response, and simple operation for applications in biological, medical, and chemical detection, but limited by the poor stability and high cost of the electrode materials. In this work, we used PtNi lagged-like nanowire
Hyun-Uk Park et al.
Ultrasonics sonochemistry, 58, 104673-104673 (2019-09-27)
In this work, we introduce composition-tunable core-shell-like PdM@Pt (M = Mn and Fe) nanoparticles (NPs) on carbon support (PdM@Pt/C) synthesized by one-pot sonochemical reactions using high-intensity ultrasonic probe (150 W, 20 kHz, with 13 mm solid probe) and investigate their electrocatalytic performance for oxygen reduction

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service