Skip to Content
Merck
All Photos(2)

Key Documents

278343

Sigma-Aldrich

1,2,4,5-Tetrabromobenzene

97%

Synonym(s):

2,3,5,6-Tetrabromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Br4
CAS Number:
Molecular Weight:
393.70
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

mp

180-182 °C (lit.)

functional group

bromo

SMILES string

Brc1cc(Br)c(Br)cc1Br

InChI

1S/C6H2Br4/c7-3-1-4(8)6(10)2-5(3)9/h1-2H

InChI key

QCKHVNQHBOGZER-UHFFFAOYSA-N

General description

The single structure of 1,2,4,5-tetrabromobenzene as actuating elements was studied and the relations between their mechanical properties and kinematic profile were determined.

Application

1,2,4,5-Tetrabromobenzene has been used in the synthesis of diethynyltriptycene-linked dipyridyl ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manas K Panda et al.
Angewandte Chemie (International ed. in English), 56(28), 8104-8109 (2017-05-26)
In salient effects, still crystals of solids that switch between phases acquire a momentum and are autonomously propelled because of rapid release of elastic energy accrued during a latent structural transition induced by heat, light, or mechanical stimulation. When mechanical
Mónica Montory et al.
Environment international, 142, 105803-105803 (2020-06-21)
Biotransport is often associated with migration patterns of species, including large, anadromous salmonids. Several studies have reported biotransport of persistent organic pollutants in the Northern Hemisphere, but there is no published information on biotransport ocurring south of the equator. Chile's
Eva Junqué et al.
Environmental research, 156, 714-724 (2017-05-10)
In this research the levels of organochlorine compounds (OCs) and mercury (Hg) in several food items from Menorca Island were presented. The dietary exposure assessment was performed in children population from the island. Finally, body burden of OCs and Hg
Subash Chandra Sahoo et al.
Journal of the American Chemical Society, 135(37), 13843-13850 (2013-07-31)
A paradigm shift from hard to flexible, organic-based optoelectronics requires fast and reversible mechanical response from actuating materials that are used for conversion of heat or light into mechanical motion. As the limits in the response times of polymer-based actuating
Jeremy J Kodanko et al.
Organic letters, 7(21), 4585-4588 (2005-10-08)
[reaction: see text] An efficient route to a new family of dinucleating ligands has been developed. A convergent strategy to these ligands involved dual Sonogashira cross-coupling of 2,3-diethynyltriptycene with a variety of functionally diverse 5-bromopyridines. The resultant ligands were accessed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service