Skip to Content
Merck
All Photos(1)

Key Documents

131857

Sigma-Aldrich

1,2,4,5-Tetrachlorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl4
CAS Number:
Molecular Weight:
215.89
Beilstein:
1618315
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

chips

bp

240-246 °C (lit.)

mp

138-140 °C (lit.)

SMILES string

Clc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C6H2Cl4/c7-3-1-4(8)6(10)2-5(3)9/h1-2H

InChI key

JHBKHLUZVFWLAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,4,5-Tetrachlorobenzene was used to study the environmental behavior of hexachlorobutadiene (HCBD).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiyan Zhang et al.
Environmental science & technology, 48(3), 1525-1531 (2014-01-10)
Although hexachlorobutadiene (HCBD) was recently proposed as a candidate persistent organic pollutant (POP) under the Stockholm Convention, information about its environmental levels and distributions is still very limited. In this work, HCBD was determined in the sewage sludge from 37
Hun-Young Wee et al.
Journal of hazardous materials, 155(1-2), 1-9 (2007-12-07)
Palladium-catalyzed hydrodehalogenation (HDH) was applied for destroying 1,2,4,5-tetrachlorobenzene (TeCB) in mixtures of water and ethanol. This investigation was performed as a critical step in the development of a new technology for clean-up of soil contaminated by halogenated hydrophobic organic contaminants.
Oxygen consumption of juvenile rainbow trout (Oncorhynchus mykiss) exposed to sublethal concentrations of 1,2,4,5-tetrachlorobenzene and tetrachloroguaiacol.
R Yang et al.
Bulletin of environmental contamination and toxicology, 59(3), 479-485 (1997-09-01)
S Beil et al.
European journal of biochemistry, 247(1), 190-199 (1997-07-01)
The bacterium, Burkholderia (previously Pseudomonas) sp. strain PS12, reported earlier to degrade 1,2,4-trichlorobenzene is shown here to utilize also 1,2,4,5-tetrachlorobenzene (Cl4-benzene) as a growth substrate. To investigate the possibility that this organism attacks Cl4-benzene with a chlorobenzene dioxygenase which concomitantly
D L Gustafson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 53(2), 245-252 (2000-03-04)
Of the twelve different chlorobenzene isomers, a thorough evaluation of carcinogenicity has only been assessed on monochlorobenzene, 1,2-, and 1,4-dichlorobenzene, and hexachlorobenzene. In the studies presented here, we measured the ability of 1,4-dichlorobenzene (DCB), 1,2,4,5-tetrachlorobenzene (TeCB), pentachlorobenzene (PeCB), and hexachlorobenzene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service