Skip to Content
Merck
All Photos(1)

Key Documents

277231

Sigma-Aldrich

Phenyltin trichloride

98%

Synonym(s):

Trichlorophenylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SnCl3
CAS Number:
Molecular Weight:
302.17
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.585 (lit.)

bp

142-143 °C/25 mmHg (lit.)

density

1.839 g/mL at 25 °C (lit.)

SMILES string

Cl[Sn](Cl)(Cl)c1ccccc1

InChI

1S/C6H5.3ClH.Sn/c1-2-4-6-5-3-1;;;;/h1-5H;3*1H;/q;;;;+3/p-3

InChI key

UBOGEXSQACVGEC-UHFFFAOYSA-K

Application

Phenyltin trichloride can be used:
  • In the preparation of organotin(IV) complexes as potent antimicrobial agents.
  • As a reactant in the Stille coupling reaction catalyzed by peptide-templated Pd nanoparticles.
  • As a tin (Sn) source in the synthesis of tin-doped TiO2 photoanodes applicable in dye-sensitized solar cells.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Peptide template effects for the synthesis and catalytic application of Pd nanoparticle networks
Jakhmola A, et al.
Journal of Materials Chemistry, 20(8), 1522-1531 (2010)
Organotin (IV) complexes with 2-acetylpyridine benzoyl hydrazones: antimicrobial activity
Despaigne Angel AR, et al.
Journal of the Brazilian Chemical Society, 21(7), 1247-1257 (2010)
Ji-Liang Zhang et al.
Ecotoxicology and environmental safety, 169, 76-84 (2018-11-14)
The impacts of triphenyltin (TPT) on ecological health have been of great concern due to their widespread use and ubiquity in aquatic ecosystems. However, little is known about the effects of TPT on the reproductive behaviors of fishes. Therefore, the
Influence of Sn source on the performance of dye-sensitized solar cells based on Sn-doped TiO2 photoanodes: A strategy for choosing an appropriate doping source
Duan Y, et al.
Electrochimica Acta, 107(8), 473-480 (2013)
Nguyen Hoang Lam et al.
Marine pollution bulletin, 121(1-2), 302-312 (2017-06-18)
A simultaneous monitoring study on organotins (butyltins and phenyltins) and most frequently used alternative antifouling biocides (Irgarol 1051, Diuron, Sea-Nine 211 and M1) in water and sediments (n=44) collected from three Special Management Sea Areas operated by Korean government. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service