Skip to Content
Merck
All Photos(1)

Documents

262919

Sigma-Aldrich

Tantalum

foil, thickness 0.025 mm, ≥99.9% trace metals basis

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
EC Number:
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Quality Level

Assay

≥99.9% trace metals basis

form

foil

autoignition temp.

572 °F

resistivity

13.5 μΩ-cm, 20°C

thickness

0.025 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

The structure of tantalum is body centered cubic (BCC) metal. It is a refractory metal; stable up to very high temperatures. Tantalum is ductile at room temperature and moderately ductile at cryogenic temperatures.

Quantity

150 × 150 mm (appoximately 9 g)
150 × 500 mm (approximately 31 g)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Morphology of Tensile Failure in Tantalum
Boyce BL, et al.
Metallurgical and Materials Transactions A: Physical Metallurgy and Materials Science, 44(10), 4567-4580 (2013)
Orbital ultrasound in the selective screening of a dural-cavernous sinus fistula.
Hamid Shokoohi et al.
European journal of emergency medicine : official journal of the European Society for Emergency Medicine, 20(2), 137-139 (2013-02-26)
Scott M Sporer et al.
Clinical orthopaedics and related research, 470(11), 3156-3163 (2012-09-25)
Stabilization of a pelvic discontinuity with a posterior column plate with or without an associated acetabular cage sometimes results in persistent micromotion across the discontinuity with late fatigue failure and component loosening. Acetabular distraction offers an alternative technique for reconstruction
S Nag et al.
Journal of the mechanical behavior of biomedical materials, 16, 21-28 (2012-11-10)
Microstructure and mechanical properties of laser deposited complex quaternary Ti-34Nb-7Zr-7Ta (all wt%), an orthopedic load-bearing implant alloy, has been investigated in detail in both as-deposited as well as heat-treated (β-solutionized and quenched) conditions. The difference in stress-strain behavior of this
Thomas Larzon et al.
Vascular, 21(2), 87-91 (2013-03-20)
Control of back bleeding from the hypogastric artery into the aneurysm after endovascular aneurysm repair (EVAR) of a ruptured aorto-iliac aneurysm may be necessary in order to avoid a type II endoleak. It is an emergency situation and selective catheterization

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service