Skip to Content
Merck
All Photos(1)

Documents

357243

Sigma-Aldrich

Tantalum

foil, thickness 0.05 mm, ≥99.9% trace metals basis

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
EC Number:
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Quality Level

Assay

≥99.9% trace metals basis

form

foil

autoignition temp.

572 °F

resistivity

13.5 μΩ-cm, 20°C

thickness

0.05 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

The structure of tantalum is body centered cubic (BCC) metal. It is a refractory metal; stable up to very high temperatures. Tantalum is ductile at room temperature and moderately ductile at cryogenic temperatures. Compton profile of highly pure thin elemental foil of tantalum was investigated using IGP type coaxial photon detector.1

Application

0.05mm thick tantalum foil was filled with 12C or 13C powder to act as carbon source emitting low energy carbon atoms.

Quantity

  • 100×100 mm (approximately 8.4 g)
  • 150×150 mm (approximately 18.9 g)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple and clean source of low energy atomic carbon
Krasnokutski SA and Huisken F
Applied Physics Letters, 105 (2014)
The Morphology of Tensile Failure in Tantalum
Boyce BL, et al.
Metallurgical and Materials Transactions A: Physical Metallurgy and Materials Science, 44(10), 4567-4580 (2013)
Thomas Larzon et al.
Vascular, 21(2), 87-91 (2013-03-20)
Control of back bleeding from the hypogastric artery into the aneurysm after endovascular aneurysm repair (EVAR) of a ruptured aorto-iliac aneurysm may be necessary in order to avoid a type II endoleak. It is an emergency situation and selective catheterization
Orbital ultrasound in the selective screening of a dural-cavernous sinus fistula.
Hamid Shokoohi et al.
European journal of emergency medicine : official journal of the European Society for Emergency Medicine, 20(2), 137-139 (2013-02-26)
Yongyuan Guo et al.
International journal of molecular medicine, 31(3), 689-697 (2013-01-23)
Titanium (Ti) alloys of the β-type are highly attractive metallic materials for biomedical applications due to their low elastic modulus, high corrosion resistance and notable biocompatibility. A new β-type Ti35Nb2Ta3Zr alloy with a low Young's modulus of approximately 48 GPa was previously

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service