Skip to Content
Merck
All Photos(1)

Key Documents

103543

Sigma-Aldrich

4-Nitroanisole

97%

Synonym(s):

1-Methoxy-4-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OCH3
CAS Number:
Molecular Weight:
153.14
Beilstein:
1865361
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39032065
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

density

1.233 g/mL at 25 °C (lit.)

functional group

nitro

SMILES string

COc1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H7NO3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3

InChI key

BNUHAJGCKIQFGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Nitroanisole undergoes photochemical nucleophilic aromatic substitution by hydroxide ion to form 4-methoxyphenol and 4-nitrophenol.

Application

4-Nitroanisole was used as probe to determine Π* of Kamlet-Taft solvent parameters and high pressure and supercritical water in the temperature range of 16-420 °C. 4-Nitroanisole was used as carbon and energy supplement for the isolation of Rhodococcus strains.

Biochem/physiol Actions

4-Nitroanisole is O-demethylated to 4-nitrophenol by human liver microsomes.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petr Klán et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 1(12), 1012-1016 (2003-03-29)
A temperature-sensitive photochemical nucleophilic aromatic substitution on 4-nitroanisole by a hydroxide ion in homogeneous solutions, in a two-phase system under phase-transfer catalysis conditions, and in the microwave field is reported. It was found that reaction regioselectivity dramatically changes with temperature
D J Sequeira et al.
Biochimica et biophysica acta, 1207(2), 179-186 (1994-08-17)
The goal of this study was to examine the effect of duration of ethylbenzene exposure on cytochrome P-450-dependent activities. Male rats were treated with ethylbenzene by intraperitoneal injection for either 1 or 3 days, and microsomal preparations were examined for
Andrea Ciani et al.
Chemosphere, 61(10), 1410-1418 (2005-07-12)
The kinetics of direct photochemical transformations of organic compounds in light absorbing and scattering media has been sparsely investigated. This is mostly due to the experimental difficulties to assess the major parameters: light intensity in porous media, the reaction quantum
A Schäfer et al.
Biodegradation, 7(3), 249-255 (1996-06-01)
Two Rhodococcus strains, R. opacus strain AS2 and R. erythropolis strain AS3, that were able to use 4-nitroanisole as the sole source of carbon and energy, were isolated from environmental samples. The first step of the degradation involved the O-demethylation
Vadim Samoilov et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-15)
In this study approaches for chemical conversions of the renewable compounds 1,2-propanediol (1,2-PD) and 2,3-butanediol (2,3-BD) that yield the corresponding cyclic ketals and glycol ethers have been investigated experimentally. The characterization of the obtained products as potential green solvents and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service