Skip to Content
Merck
All Photos(3)

Documents

233749

Sigma-Aldrich

Calcium bromide hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaBr2 · xH2O
CAS Number:
Molecular Weight:
199.89 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

98%

form

powder or chunks

composition

Degree of hydration, ≤1

pH

5 (20 °C, 50 g/L)

SMILES string

[H]O[H].Br[Ca]Br

InChI

1S/2BrH.Ca.H2O/h2*1H;;1H2/q;;+2;/p-2

InChI key

NQMKEZBQFUTOQC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium bromide hydrate may be used in the preparation of calcium-carbohydrate complexes to study the structural factors that influence the binding interactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium binding to carbohydrates: crystal structure of a hydrated calcium bromide salt of lactobionic acid.
Cook WJ & Bugg CE
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 29(2), 215-222 (1973)
Calcium binding to carbohydrates. Crystal structure of a hydrated calcium bromide complex of lactose.
C E Bugg
Journal of the American Chemical Society, 95(3), 908-913 (1973-02-07)
L DeLucas et al.
Carbohydrate research, 41, 18-29 (1975-05-01)
Three-dimensional X-ray diffraction data were used to determine the crystal structure of alpha-D-glucuronate CaBr times 3H20, a model system for investigating the factors involved in the binding of calcium ions to D-glucuronate residues of oligo-and poly-saccharides. Crystals of the salt
Calcium binding to galactose. Crystal structure of a hydrated a-galactose-calcium bromide complex.
Cook WJ and Bugg CE.
Journal of the American Chemical Society, 95(19), 6442-6446 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service