Skip to Content
Merck
All Photos(1)

Documents

213314

Sigma-Aldrich

Antimony(III) ethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb(OC2H5)3
CAS Number:
Molecular Weight:
256.94
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

liquid

composition

Sb, 46.6-48.1% iodometric
Sb, 46.6-48.1% titration (by iodine)

reaction suitability

core: antimony
reagent type: catalyst

refractive index

n20/D 1.495 (lit.)

density

1.513 g/mL at 25 °C (lit.)

SMILES string

CCO[Sb](OCC)OCC

InChI

1S/3C2H5O.Sb/c3*1-2-3;/h3*2H2,1H3;/q3*-1;+3

InChI key

JGOJQVLHSPGMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinchun Qiu et al.
The journal of physical chemistry. A, 120(46), 9257-9269 (2016-10-22)
Density functional theory calculations were used to design an anion receptor that utilizes antimony(III) secondary bonding interactions. Calculations were performed on promising motifs found in the chemical literature where two antimony sites were found in close proximity to a halide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service