Skip to Content
Merck
All Photos(1)

Key Documents

21185

Sigma-Aldrich

Calcium L-lactate hydrate

≥98.0% (calc. based on dry substance, KT)

Synonym(s):

(S)-(+)-2-Hydroxypropanoic acid hemicalcium salt, L-Lactic acid hemicalcium salt, Sarcolactic acid hemicalcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3CH(OH)COO]2Ca · xH2O
CAS Number:
Molecular Weight:
218.22 (anhydrous basis)
Beilstein:
4923285
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (calc. based on dry substance, KT)

form

solid

optical activity

[α]20/D −4.2±0.5°, c = 5% in H2O

impurities

~4 mol/mol water

SMILES string

O.C[C@H](O)C(=O)O[Ca]OC(=O)[C@H](C)O

InChI

1S/2C3H6O3.Ca.H2O/c2*1-2(4)3(5)6;;/h2*2,4H,1H3,(H,5,6);;1H2/q;;+2;/p-2/t2*2-;;/m00../s1

InChI key

MDAVASCOAJMZHZ-NVKWYWNSSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium L-lactate hydrate may be used as a calcium precursor in the preparation of calcium-oxide based sorbents for CO2 capture.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancing the performance of CaO/CuO based composite for CO2 capture in a combined Ca-Cu chemical looping process.
Qin C, et al.
Chemical Engineering Journal, 228, 75-86 (2013)
Synthesis of sintering-resistant sorbents for CO2 capture.
Liu W, et al.
Environmental Science & Technology, 44(8), 3093-3097 (2010)
Calcium precursors for the production of CaO sorbents for multicycle CO2 capture.
Liu W, et al.
Environmental Science & Technology, 44(2), 841-847 (2009)
Michael T Lam et al.
The Journal of experimental medicine, 216(12), 2778-2799 (2019-10-12)
Hemophagocytic lymphohistiocytosis (HLH) is characterized by immune dysregulation due to inadequate restraint of overactivated immune cells and is associated with a variable clinical spectrum having overlap with more common pathophysiologies. HLH is difficult to diagnose and can be part of
Nevena Grdović et al.
PloS one, 9(7), e101172-e101172 (2014-07-06)
The diabetes prevention paradigm envisages the application of strategies that support the maintenance of appropriate β-cell numbers. Herein we show that overexpression of CXC chemokine ligand12 (CXCL12) considerably improves the viability of isolated rat Langerhans islet cells and Rin-5F pancreatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service