Skip to Content
Merck
All Photos(1)

Documents

211729

Sigma-Aldrich

Chlorotrimethylgermane

98%

Synonym(s):

Trimethylchlorogermane, Trimethylgermanium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3GeCl
CAS Number:
Molecular Weight:
153.20
Beilstein:
3535109
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.433 (lit.)

bp

102 °C (lit.)

mp

−13 °C (lit.)

density

1.24 g/mL at 25 °C (lit.)

SMILES string

C[Ge](C)(C)Cl

InChI

1S/C3H9ClGe/c1-5(2,3)4/h1-3H3

InChI key

ZZBNZZCHSNOXOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Virginia Solar Fernandez et al.
Antioxidants & redox signaling, 32(4), 217-227 (2019-11-07)
Aims: Nuclear factor (erythroid-derived 2)-like-2 factor (NRF-2) is a transcription factor well known to provide an advantage for cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service