Skip to Content
Merck
All Photos(3)

Key Documents

204455

Sigma-Aldrich

Strontium carbonate

99.995% trace metals basis

Synonym(s):

Strontianite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCO3
CAS Number:
Molecular Weight:
147.63
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38020224
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks
solubility:
dilute aqueous acid: slightly soluble(lit.)

grade

for analytical purposes

Assay

99.995% trace metals basis

form

powder and chunks

impurities

≤55.0 ppm Trace Metal Analysis

mp

1494 °C (lit.)

solubility

dilute aqueous acid: slightly soluble(lit.)

density

3.7 g/mL at 25 °C (lit.)

SMILES string

[Sr++].[O-]C([O-])=O

InChI

1S/CH2O3.Sr/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

LEDMRZGFZIAGGB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Strontium carbonate: Extensively used in the production of glazes in ceramics, where it acts as a flux and introduces strontium into the glaze for improved finish and durability. It is also crucial in the manufacture of ferrite magnets, providing the necessary strontium content to enhance magnetic properties. Additionally, its role in pyrotechnics is to contribute to the vibrant red hues seen in fireworks displays (Sigma-Aldrich, CAS 1633-05-2).

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geoffrey M Bowers et al.
Solid state nuclear magnetic resonance, 29(1-3), 95-103 (2005-09-20)
The only stable NMR-active isotope of strontium, (87)Sr, is a spin-9/2 quadrupolar nucleus that has a low gyromagnetic ratio, a low natural abundance, and a large nuclear electric quadrupole moment. In this work, we utilize the quadrupolar Carr-Purcell-Meiboom-Gill (QCPMG) pulse
Wen-Yu Qian et al.
International journal of nanomedicine, 7, 5781-5792 (2012-11-28)
Strontium carbonate nanoparticles (SCNs), a novel biodegradable nanosystem for the pH-sensitive release of anticancer drugs, were developed via a facile mixed solvent method aimed at creating smart drug delivery in acidic conditions, particularly in tumor environments. Structural characterization of SCNs
Sunkyung Choi et al.
Environmental science & technology, 40(8), 2608-2614 (2006-05-11)
Radioactive strontium (90Sr) is an important constituent of the complex wastes from past nuclear weapons production and has been stored in underground tanks at U.S. DOE sites (e.g., Hanford, WA). Using bulk and microfocused EXAFS spectroscopy, we examined temporal changes
Akiko Sakai et al.
Dental materials journal, 31(2), 197-205 (2012-03-27)
Sr-containing carbonate apatite (SrCAp) specimens of varied Sr contents, ranging from 0 to 13.3 mol%, were prepared through a phosphate treatment of set gypsum-and-carbonate mixture at 100°C for 7 days. Effects of Sr content in SrCAp on microstructure, osteoblast-like cell
R B Greegor et al.
Science (New York, N.Y.), 275(5305), 1452-1454 (1997-03-07)
An x-ray spectroscopic study of scleractinian coral skeletons indicated that, although some strontium substitutes for calcium in the aragonite structure, at concentrations of about 7500 parts per million, as much as 40 percent of the strontium resides in strontianite (SrCO3).

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service