Skip to Content
Merck
All Photos(1)

Key Documents

450812

Sigma-Aldrich

Copper(II) oxide

powder, 99.99% trace metals basis

Synonym(s):

Cupric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder

application(s)

battery manufacturing

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Copper(II)oxide or cupric oxide (CuO) is a chemically stable black solid. As a mineral, it is called tenorite and is a critical product of copper mining. Our CuO with a trace metals purity of 99.99% is a high purity material ideal for applications in electronics, ceramics, and catalysis. CuO is a medium bandgap semiconductor with a bandgap in the range of 1.2-2.6 eV depending on the preparation conditions and usually shows p-type conductivity due to the presence of copper vacancy defects. In ceramic sciences, CuO is a key componentof many types of high Tc superconducting materials like BaYCu3O9-δ and Bi2Sr2CuO6+δ; the CuO is usually milled or mixed with other metal oxides and sintered at high temperatures to obtain complex metal oxides. In the field of catalysis, CuO is also widely used as a precursor to synthesize electrocatalysts for organic synthesis and degradation of organic pollutants.

Application

  • Copper(II) Oxide Nanoparticles Embedded within a PEDOT Matrix for Hydrogen Peroxide Electrochemical Sensing.: This study explores the use of copper(II) oxide nanoparticles embedded in a PEDOT matrix for effective hydrogen peroxide electrochemical sensing, highlighting the material′s potential in energy storage applications (Lete et al., 2022).
  • Green Fabrication of Nonenzymatic Glucose Sensor Using Multi-Walled Carbon Nanotubes Decorated with Copper (II) Oxide Nanoparticles for Tear Fluid Analysis.: Researchers developed a nonenzymatic glucose sensor using copper(II) oxide nanoparticles on multi-walled carbon nanotubes, showcasing copper oxide′s versatility in high-capacity energy storage solutions and renewable energy applications (Asgari Kheirabadi et al., 2022).
  • Facile Synthesis of Copper(II) Oxide Nanospheres Covered on Functionalized Multiwalled Carbon Nanotubes Modified Electrode as Rapid Electrochemical Sensing Platform for Super-Sensitive Detection of Antibiotic.: This paper discusses the synthesis of copper(II) oxide nanospheres on multiwalled carbon nanotubes, emphasizing their application in rapid and sensitive electrochemical sensing, relevant to battery technology (Chen et al., 2019).
  • Composite Planar Electrode for Sensing Electrochemical Oxygen Demand.: This study focuses on the development of a composite planar electrode incorporating copper(II) oxide for sensing electrochemical oxygen demand, relevant to improving battery efficiency (Orozco et al., 2008).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumanta Kumar Meher et al.
Nanoscale, 5(5), 2089-2099 (2013-02-06)
In the quest to enhance the selectivity and sensitivity of novel structured metal oxides for electrochemical non-enzymatic sensing of glucose, we report here a green synthesis of unique sandwich-structured CuO on a large scale under microwave mediated homogeneous precipitation conditions.
Guangliang Cui et al.
Scientific reports, 3, 1250-1250 (2013-02-15)
Heterostructure material that acts as resonant tunneling system is a major scientific challenge in applied physics. Herein, we report a resonant tunneling system, quasi-2D Cu(2)O/SnO(2) p-n heterostructure multi-layer film, prepared by electrochemical deposition in a quasi-2D ultra-thin liquid layer. By
Lisha Zhou et al.
Nanoscale, 5(4), 1564-1569 (2013-01-18)
Stable Cu(2)O nanocrystals of around 3 nm were uniformly and densely grown on functionalized graphene sheets (FGS), which act as molecular templates instead of surfactants for controlled nucleation; the distribution density of nanocrystals can be easily controlled by FGS with
Wei Jian Foo et al.
Nanoscale, 5(2), 759-764 (2012-12-12)
Here we have demonstrated the preparation of high-quality, monodispersed and tunable phases of Cu nanoparticles. Structural and chemical composition studies depict the evolution of Cu-Cu(2)O-CuO nanoparticles at various process stages. The loading of Cu and Cu oxide nanoparticles on TiO(2)
Yifu Yu et al.
Nanoscale, 5(3), 904-909 (2012-12-21)
A simple and feasible contact mode called gravitational contact mode (GCM) was developed for the first time to imitate the practical state between soot and catalyst. By simulating rainwater adsorption on a lawn in nature, we synthesized a lawn-like CuO

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service