Skip to Content
Merck
All Photos(1)

Key Documents

194557

Sigma-Aldrich

Methoxyacetic acid

98%

Synonym(s):

NSC 7300

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2COOH
CAS Number:
Molecular Weight:
90.08
Beilstein:
635800
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

202-204 °C (lit.)

mp

7-9 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

functional group

carboxylic acid
ether

SMILES string

COCC(O)=O

InChI

1S/C3H6O3/c1-6-2-3(4)5/h2H2,1H3,(H,4,5)

InChI key

RMIODHQZRUFFFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methoxyacetic acid is the major metabolite of ester phthalates widely used in industry as gelling, viscosity and stabilizer reagents.

Application

Methoxyacetic acid was used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis in ovo.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Yang et al.
Urology, 81(3), 696-696 (2013-01-08)
To investigate the expression pattern of Ras-related protein 1 (Rap1) during testicular development and to clarify whether its expression is developmentally regulated and whether this expression is involved in the process of germ cell apoptosis. The expression pattern of Rap1
Gargi Bagchi et al.
Reproductive biology and endocrinology : RB&E, 9, 42-42 (2011-04-02)
Optimal androgen signaling is critical for testicular development and spermatogenesis. Methoxyacetic acid (MAA), the primary active metabolite of the industrial chemical ethylene glycol monomethyl ether, disrupts spermatogenesis and causes testicular atrophy. Transcriptional trans-activation studies have indicated that MAA can enhance
Bao-Lin Li et al.
Chirality, 20(1), 35-39 (2007-10-26)
The resolutions of (9-anthryl)methoxyacetic acid (9AMAA) and (9-anthryl)hydroxyacetic acid (9AHAA) were performed by capillary electrophoresis using hydroxypropyl-beta-cyclodextrin (HP-beta-CD) as a chiral selector. Various factors affecting migration time and resolutions of these compounds were investigated with a run voltage of 20
Didik Priyandoko et al.
PloS one, 6(5), e19552-e19552 (2011-05-17)
The present day lifestyle heavily depends on industrial chemicals in the form of agriculture, cosmetics, textiles and medical products. Since the toxicity of the industrial chemicals has been a concern to human health, the need for alternative non-toxic natural products
Joshua F Robinson et al.
Toxicological sciences : an official journal of the Society of Toxicology, 118(2), 675-685 (2010-09-25)
Rat postimplantation whole-embryo culture (WEC) is a promising alternative test for the assessment of developmental toxicity. Toxicogenomic-based approaches may improve the predictive ability of the WEC model by providing a means to identify compound-specific mechanistic responses associated with embryotoxicity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service