Skip to Content
Merck
All Photos(2)

Documents

149209

Sigma-Aldrich

Methyl methoxyacetate

99%

Synonym(s):

Methoxyacetic acid methyl ester, Methyl α-methoxyacetate, Methyl 2-methoxyacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2COOCH3
CAS Number:
Molecular Weight:
104.10
Beilstein:
1742944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.396 (lit.)

bp

129-130 °C (lit.)

density

1.051 g/mL at 25 °C (lit.)

SMILES string

COCC(=O)OC

InChI

1S/C4H8O3/c1-6-3-4(5)7-2/h3H2,1-2H3

InChI key

QRMHDGWGLNLHMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl methoxyacetate was used in the preparation of 4-hydroxy-2-mercapto-5-methoxypyrimidine (II).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

888. Pyrimidines. Part XI. Synthesis of 5-hydroxypyrimidine and related compounds.
Chesterfield JH, et al.
Journal of the Chemical Society, 4590-4596 (1960)
Shigehiro Namiki et al.
The International journal of neuroscience, 119(2), 214-226 (2009-01-07)
Loading neurons with membrane permeable Ca2+ indicators is a core experimental procedure in functional multineuron Ca2+ imaging (fMCI), an optical technique for monitoring multiple neuronal activities. Although fMCI has been applied to several brain networks, including cerebral cortex, hippocampus, and
[Information from the Soviet Toxicology Center].
Farmakologiia i toksikologiia, 43(6), 750-751 (1980-11-01)
K Thompson et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 87(7), 468-483 (2012-09-19)
Stimulated by difficulties experienced when using fluo-3 AM, we developed a general mechanistic model to aid understanding and practical application of calcium probes applied as acetoxymethyl (AM) esters. Several practical issues previously overlooked or under-emphasized are considered by this model.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service