Skip to Content
Merck
All Photos(4)

Key Documents

191884

Sigma-Aldrich

Hydroxypropyl cellulose

average Mw ~100,000, powder, 20 mesh particle size (99% through)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

biological oxygen demand (BOD) 14,000 ppm

Quality Level

form

powder

autoignition temp.

752 °F

mol wt

average Mw ~100,000

impurities

<5 wt. %

particle size

20 mesh (99% through)

pH

5.0-8.5

interfacial tension mineral oil

12.5 dyn/cm, 0.1 wt. % in H2O (vs. mineral oil)

viscosity

65-175 cP, 5 wt. % in H2O(25 °C, Brookfield, spindle #1) (30 rpm)(lit.)

solubility

polar organic solvents: soluble

density

0.5 g/mL at 25 °C (lit.)

SMILES string

N(C)(C)c1cc(c(cc1)C(N)CC)C

InChI

1S/C12H20N2/c1-5-12(13)11-7-6-10(14(3)4)8-9(11)2/h6-8,12H,5,13H2,1-4H3

InChI key

RRHXDYJWVYFMKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxypropyl cellulose (HPC) is a neutral polysaccharide prepared by reacting propylene oxide with alkali cellulose at high temperatures and pressure. It is soluble in water and many polar organic solvents. HPC is widely used as a food additive and in drug delivery systems.

Application

Hydroxypropyl cellulose can be used in the following applications:
  • HPC is soluble in organic solvents which makes it suitable for the preparation of polymer gel electrolytes for electrochemical devices. The network-forming ability of HPC can improve the mechanical and electrical properties of gel electrolytes.
  • Electrospun hydroxypropyl cellulose nanofibres can be used to develop drug delivery systems for a variety of hydrophobic drugs.
  • Hydroxypropylcellulose combined with an effervescent agent like sodium carbonate can be usedas a binder and functional material to prepare floating tablets for thecontrolled release of ofloxacin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeremiah F Kelleher et al.
AAPS PharmSciTech, 21(1), 23-23 (2019-12-14)
The development of oral solid dosage forms, such as tablets that contain a high dose of drug(s), requires polymers and other additives to be incorporated at low levels as possible, to keep the final tablet weight low, and, correspondingly, the
Xiaole Qi et al.
International journal of pharmaceutics, 489(1-2), 210-217 (2015-05-10)
To prolong the residence time of dosage forms within gastrointestinal trace until all drug released at desired rate was one of the real challenges for oral controlled-release drug delivery system. Herein, we developed a fine floating tablet via compression coating
Daniela M Correia et al.
Materials (Basel, Switzerland), 13(10) (2020-05-21)
This study reports a versatile method for the development of cellulose nanocrystals (CNCs) and water-soluble cellulose derivatives (methyl cellulose (MC), hydroxypropyl cellulose (HPC), and sodium carboxymethyl cellulose (NaCMC)) films comprising the ionic liquid (IL) 2-hydroxy-ethyl-trimethylammonium dihydrogen phosphate ([Ch][DHP]) for actuator
Jianjun Yang et al.
International orthopaedics, 37(3), 515-521 (2012-11-28)
The aim of this study was to determine whether hydroxypropylcellulose (HPC) coating of polyethylene terephthalate (PET) artificial ligaments enhances graft osseointegration in the bone tunnel. Thirty New Zealand white rabbits underwent artificial ligament graft transplantation in the bilateral proximal tibia
Maya Fujiwara et al.
Chemical & pharmaceutical bulletin, 60(11), 1380-1386 (2012-11-06)
We evaluated models for predicting spray mist diameter suitable for scaling-up the fluidized bed granulation process. By precise selection of experimental conditions, we were able to identify a suitable prediction model that considers changes in binder solution, nozzle dimension, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service