All Photos(1)
About This Item
Linear Formula:
CH3O2CCH2CH(CH3)CO2CH3
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.42 (lit.)
bp
196 °C (lit.)
density
1.076 g/mL at 25 °C (lit.)
functional group
ester
SMILES string
COC(=O)CC(C)C(=O)OC
InChI
1S/C7H12O4/c1-5(7(9)11-3)4-6(8)10-2/h5H,4H2,1-3H3
InChI key
NFOQJNGQQXICBY-UHFFFAOYSA-N
Related Categories
Application
Dimethyl methylsuccinate was used in the synthesis of β-half esters.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
181.4 °F - closed cup
Flash Point(C)
83 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
El-Abbady AM, et al.
Canadian Journal of Chemistry, 43(4), 928-934 (1965)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service