Skip to Content
Merck
All Photos(1)

Documents

73605

Supelco

Dimethyl succinate

analytical standard

Synonym(s):

Succinic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2CH2COOCH3
CAS Number:
Molecular Weight:
146.14
Beilstein:
956776
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.3 mmHg ( 20 °C)

Assay

≥97.0% (GC)

autoignition temp.

689 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.418-1.420
n20/D 1.419 (lit.)

bp

200 °C (lit.)

mp

16-19 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COC(=O)CCC(=O)OC

InChI

1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3

InChI key

MUXOBHXGJLMRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cinzia Donato et al.
Cell reports, 32(10), 108105-108105 (2020-09-10)
Circulating tumor cells (CTCs) are shed from solid cancers in the form of single or clustered cells, and the latter display an extraordinary ability to initiate metastasis. Yet, the biological phenomena that trigger the shedding of CTC clusters from a
Ayesha B Alvero et al.
Oncotarget, 5(18), 8703-8715 (2014-09-23)
Survival rate in ovarian cancer has not improved since chemotherapy was introduced a few decades ago. The dismal prognosis is mostly due to disease recurrence where majority of the patients succumb to the disease. The demonstration that tumors are comprised
Danielle B Ulanet et al.
PloS one, 9(12), e115144-e115144 (2014-12-17)
Recent work has highlighted glutaminase (GLS) as a key player in cancer cell metabolism, providing glutamine-derived carbon and nitrogen to pathways that support proliferation. There is significant interest in targeting GLS for cancer therapy, although the gene is not known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service