Skip to Content
Merck
All Photos(3)

Key Documents

178012

Sigma-Aldrich

(1,2-Dibromoethyl)benzene

≥98%

Synonym(s):

1,2-Dibromo-1-phenylethane, Phenylethylene bromide, Styrene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(Br)CH2Br
CAS Number:
Molecular Weight:
263.96
Beilstein:
907323
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

solid

bp

139-141 °C/15 mmHg (lit.)

mp

70-74 °C (lit.)

functional group

bromo
phenyl

SMILES string

BrCC(Br)c1ccccc1

InChI

1S/C8H8Br2/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

SHKKTLSDGJRCTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The metabolism of 1,2-dibromo-1-phenylethane in rat was examined.

Application

  • Effects of the biocide methylisothiazolinone on Xenopus laevis wound healing and tail regeneration.: The study assesses the impact of 2-Methyl-4-isothiazolin-3-one on regenerative processes in amphibians, contributing to the understanding of its biological effects and potential toxicity (Delos Santos et al., 2016).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Zoetemelk et al.
Drug metabolism and disposition: the biological fate of chemicals, 15(3), 418-425 (1987-05-01)
The metabolism of 1,2-dibromo-1-phenylethane (DBPE) was studied in rats. Administration of DBPE orally, in doses of 0.25-1.25 mmol/kg (66-330 mg/kg), to male Wistar rats resulted in the excretion of a single mercapturic acid in urine. The methyl esters of three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service