All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H8Br2Fe
CAS Number:
Molecular Weight:
343.82
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
reaction suitability
core: iron
reagent type: catalyst
mp
52-55 °C
storage temp.
2-8°C
SMILES string
[Fe].Br[C]1[CH][CH][CH][CH]1.Br[C]2[CH][CH][CH][CH]2
InChI
1S/2C5H4Br.Fe/c2*6-5-3-1-2-4-5;/h2*1-4H;
InChI key
PPENAEPVSCFVJG-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zhiqiang Tan et al.
Polymers, 12(3) (2020-03-28)
Conjugated microporous polymers (CMPs) have conjugated skeleton and permanent porosity, and exhibit huge potential in developing novel functional materials for resolving the challenging energy and environment issues. Metal-containing CMPs often exhibited unique properties. In the present manuscript, ferrocene-based conjugated microporous
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service