Skip to Content
Merck
All Photos(1)

Documents

175838

Sigma-Aldrich

Glycine-d5

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2NCD2CO2D
CAS Number:
Molecular Weight:
80.10
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

technique(s)

NMR: suitable
bio NMR: suitable

mp

240 °C (dec.) (lit.)

mass shift

M+5

SMILES string

[2H]OC(=O)C([2H])([2H])N([2H])[2H]

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i1D2/hD3

InChI key

DHMQDGOQFOQNFH-LGLHGEJLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maxwell Zimmerley et al.
Journal of biomedical optics, 14(4), 044019-044019 (2009-09-04)
Coherent anti-Stokes Raman scattering (CARS) microscopy is used to determine the distribution and concentration of selected compounds in intact human hair. By generating images based on ratiometric CARS contrast, quantitative concentration maps of both water and externally applied d-glycine are
Howbeer Muhamadali et al.
Metabolites, 10(9) (2020-09-26)
In the past two decades, metabolomics has proved to be a valuable tool with many potential applications in different areas of science. However, there are still some challenges that need to be addressed, particularly for multicenter studies. These challenges are
Athanasios Mallouchos et al.
Foods (Basel, Switzerland), 9(4) (2020-04-15)
Gilthead sea bream (Sparus aurata) is one of the most important farmed Mediterranean fish species, and there is considerable interest for the development of suitable methods to assess its freshness. In the present work, gas chromatography-mass spectrometry-based metabolomics was employed
Annelie Abrahamsson et al.
Scientific reports, 7, 42733-42733 (2017-02-16)
Metabolic reprogramming is a hallmark of cancer. Nutrient availability in the tissue microenvironment determines cellular events and may play a role in breast carcinogenesis. High mammographic density is an independent risk factor for breast cancer. Whether nutrient availability differs in
Quan Yang et al.
Pharmaceutical research, 32(2), 445-457 (2014-08-06)
To characterise skin barrier function in vivo at two distinct anatomic sites using minimally invasive bioengineering and biophysical tools. In healthy human volunteers, the quantities of stratum corneum (SC) per unit area of skin on the forearm and forehead were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service