Merck
  • Characterisation of skin barrier function using bioengineering and biophysical techniques.
Characterisation of skin barrier function using bioengineering and biophysical techniques.
Pharmaceutical research (2014-08-06)
Quan Yang, Richard H Guy
ABSTRACT
To characterise skin barrier function in vivo at two distinct anatomic sites using minimally invasive bioengineering and biophysical tools. In healthy human volunteers, the quantities of stratum corneum (SC) per unit area of skin on the forearm and forehead were quantified by gravimetric and imaging techniques. Organisation of the SC intercellular lipids was evaluated as a function of position using attenuated total reflectance infrared spectroscopy (ATR-IR). The constituents of natural moisturising factor (NMF) were extracted from tape-stripped samples of the SC and by reverse iontophoresis; 21 components were identified and quantified by liquid chromatography with mass spectrometric detection. SC was quantified more accurately by imaging and was significantly thinner on the forehead than on the forearm. Intercellular lipids were more disordered near the skin surface at both sites; however, throughout forearm SC, the lipids were substantially better organised than those in the forehead. Compositionally, the NMF from forearm and forehead SC was similar, but the total amount extractable from the forehead was smaller. Taken together, the bioengineering and biophysical techniques employed demonstrate, in a complementary, objective and quantitative fashion, that SC barrier function on the forehead is less competent than that on the forearm.
Sigma-Aldrich
Glycine-d5, 98 atom % D
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Perfluoropentanoic acid, 97%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)