Skip to Content
Merck
All Photos(3)

Key Documents

175536

Sigma-Aldrich

4-Hydroxy-3-methoxybenzyl alcohol

98%

Synonym(s):

NSC 3993, Vanillyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH2OH
CAS Number:
Molecular Weight:
154.16
Beilstein:
1910044
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reagent type: cross-linking reagent

mp

110-117 °C (lit.)

SMILES string

COc1cc(CO)ccc1O

InChI

1S/C8H10O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-4,9-10H,5H2,1H3

InChI key

ZENOXNGFMSCLLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Su Kim et al.
Molecules (Basel, Switzerland), 16(7), 5349-5361 (2011-06-28)
Gastrodia elata Blume (GE) has long been used in oriental countries as a traditional herbal medicine to relieve symptoms associated with neurological ailments such as vertigo, general paralysis and epilepsy. In this study, we have investigated the effects of GE
Padmesh Venkitasubramanian et al.
The Journal of biological chemistry, 282(1), 478-485 (2006-11-15)
Aldehyde oxidoreductase (carboxylic acid reductase (Car)) catalyzes the magnesium-, ATP-, and NADPH-dependent reduction of carboxylic acids to their corresponding aldehydes. Heterologous expression of the car gene in Escherichia coli afforded purified recombinant enzyme with a specific activity nearly 50-fold lower
Kiki L Larsen et al.
The journal of physical chemistry. B, 115(39), 11470-11480 (2011-08-13)
Structural analysis of plant materials, i.e., lignin, cellulose, hemicellulose, etc., supports the growing interest of their uses, e.g., as sources for biofuels or materials. Lignin is a main polymer formed from three phenolic presursors, containing none, one, or two OMe
Anja Brencic et al.
Molecular microbiology, 51(4), 1103-1115 (2004-02-07)
Plant tumorigenesis by Agrobacterium tumefaciens requires approximately 20 Vir proteins, transcription of which is induced by a family of phenolic compounds released from plant wound sites. One Vir protein, VirH2, plays a role in the metabolism of at least one
Iris Brune et al.
Journal of biotechnology, 127(1), 21-33 (2006-08-08)
A 70mer oligonucleotide microarray was constructed to analyze genome-wide expression profiles of Corynebacterium jeikeium, a skin bacterium that is predominantly present in the human axilla and involved in axillary odor formation. Oligonucleotides representing 100% of the predicted coding regions of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service