Skip to Content
Merck
All Photos(3)

Key Documents

H18607

Sigma-Aldrich

2′-Hydroxyacetophenone

ReagentPlus®, 99%

Synonym(s):

2-Acetylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COCH3
CAS Number:
Molecular Weight:
136.15
Beilstein:
386123
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.7 (vs air)

vapor pressure

~0.2 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.558 (lit.)

bp

213 °C/717 mmHg (lit.)

mp

3-6 °C (lit.)

density

1.133 g/mL at 20 °C
1.131 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccccc1O

InChI

1S/C8H8O2/c1-6(9)7-4-2-3-5-8(7)10/h2-5,10H,1H3

InChI key

JECYUBVRTQDVAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

222.8 °F - closed cup

Flash Point(C)

106 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R R Zaky et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 81(1), 28-34 (2011-07-26)
Schiff base complexes of Cu(II), Ni(II) and Zn(II) with the o-hydroxyacetophenone [N-(3-hydroxy-2-naphthoyl)] hydrazone (H(2)o-HAHNH) containing N and O donor sites have been synthesized. Both ligand and its metal complexes were characterized by different physicochemical methods, elemental analysis, molar conductivity ((1)H
R R Zaky et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 683-694 (2012-08-14)
The o-Hydroxy acetophenone [N-(3-hydroxy-2-naphthoyl)] hydrazone (H(2)o-HAHNH) has been prepared and its structure is confirmed by elemental analysis, IR, (1)H NMR and (13)C NMR spectroscopy. It has been used to produce diverse complexes with Co(II), Cd(II), Hg(II) and U(VI)O(2) ions. The
José I Borrell et al.
Molecular diversity, 8(2), 147-157 (2004-06-24)
Two new solid-phase syntheses of substituted pyrazoles are described. The first includes supporting an o-hydroxyacetophenone on Merrifield resin, Vilsmeier-Haack formylation on the methyl group and cyclization with a substituted hydrazine to afford a pyrazole ring with two diversity centers. The
Shuai Chen et al.
Organic letters, 14(11), 2806-2809 (2012-05-17)
Flavin catalysts perform the first organocatalytic Dakin oxidation of electron-rich arylaldehydes to phenols under mild, basic conditions. Catechols are readily prepared, and the oxidation of 2-hydroxyacetophenone was achieved. Aerobic oxidation is displayed in the presence of Zn(0) as a reducing
Subramanya Gupta Sreerama et al.
Inorganic chemistry, 44(18), 6299-6307 (2005-08-30)
A series of dinuclear complexes of Mn(III), Fe(III), and Co(III) with two diazine Schiff bases, H2salhn and H2mesalhn, is reported. The Schiff bases are prepared by condensation reactions of hydrazine with salicylaldehyde (H2salhn) and with 2-hydroxyacetophenone (H2mesalhn) in 1:2 mol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service