Skip to Content
Merck
All Photos(1)

Documents

149454

Sigma-Aldrich

Hexaphenylbenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6(C6H5)6
CAS Number:
Molecular Weight:
534.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

>300 °C (lit.)

SMILES string

c1ccc(cc1)-c2c(-c3ccccc3)c(-c4ccccc4)c(-c5ccccc5)c(-c6ccccc6)c2-c7ccccc7

InChI

1S/C42H30/c1-7-19-31(20-8-1)37-38(32-21-9-2-10-22-32)40(34-25-13-4-14-26-34)42(36-29-17-6-18-30-36)41(35-27-15-5-16-28-35)39(37)33-23-11-3-12-24-33/h1-30H

InChI key

QBHWPVJPWQGYDS-UHFFFAOYSA-N

Application

Hexaphenylbenzene can be used as a starting material to synthesize:
  • 1,2,3,4,5,6-Hexacyclohexylcyclohexane by Pd/C catalyzed hydrogenation reaction.
  • Stable hexatrityl cations and porous organic polymers for applications in catalysis and gas storage.
  • Hexa-peri-hexabenzocoronene via one-pot substitution and oxidative cyclodehydrogenation reaction in the presence of t-BuCl/FeCl3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porous organic polymers based on propeller-like hexaphenylbenzene building units
Chen Q, et al.
Macromolecules, 44(14), 5573-5577 (2011)
From Hexaphenylbenzene to 1,2,3,4,5,6-Hexacyclohexylcyclohexane
Dillenburger M, et al.
Journal of the American Chemical Society, 142(30), 12916-12920 (2020)
Vandana Bhalla et al.
Chemical communications (Cambridge, England), 49(9), 895-897 (2012-12-19)
Fluorescent nanorods formed by self-assembly of hexaphenylbenzene derivative in the presence of cyanide ions serve as a sensitive colorimetric and fluorogenic sensor for the detection of trinitrotoluene (TNT) at the attogram (10(-18) g) level with a detection limit of 10.21
Synthesis and isolation of polytrityl cations by utilizing hexaphenylbenzene and tetraphenylmethane scaffolds
Rathore R, et al.
The Journal of Organic Chemistry, 69(5), 1524-1530 (2004)
A Practical One-Pot Synthesis of Soluble Hexa-peri-hexabenzocoronene and Isolation of Its Cation-Radical Salt
Rathore R and Burns CL
J. Org. React., 68(10), 4071-4074 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service