Skip to Content
Merck
All Photos(1)

Key Documents

147400

Sigma-Aldrich

N-Ethylacetamide

99%

Synonym(s):

Acetamidoethane, N-Acetylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC2H5
CAS Number:
Molecular Weight:
87.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.433 (lit.)

bp

90-92 °C/8 mmHg (lit.)

density

0.924 g/mL at 25 °C (lit.)

functional group

amide

SMILES string

CCNC(C)=O

InChI

1S/C4H9NO/c1-3-5-4(2)6/h3H2,1-2H3,(H,5,6)

InChI key

PMDCZENCAXMSOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Ethylacetamide is a suitable nylon model for mechanistic studies of dye fading.

Application

N-Ethylacetamide was used to investigate propylene carbonate and a mixture of N-methylformamide and N-ethylacetamide by broadband dielectric and mechanical shear spectroscopy. It was used in determination of reaction products of OH-oxidation of N-methylpyrrolidone under atmospheric conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphaela Kannengießer et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 16(9), 1906-1911 (2015-04-29)
The gas-phase structures and parameters describing acetyl methyl torsion of N-ethylacetamide are determined with high accuracy, using a combination of molecular beam Fourier-transform microwave spectroscopy and quantum chemical calculations. Conformational studies at the MP2 level of theory yield four minima
R Buchet et al.
Biophysical chemistry, 22(4), 249-254 (1985-10-01)
It is shown that a striking parallelism exists between the anesthetic potency of general halocarbon anesthetics and their influence on the hydrogen bond association constants in N-H...O=C type hydrogen bonds, important for shaping the ion channels. It is further shown
Tetsuya Tatsukawa et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 26(18), 4820-4825 (2006-05-05)
AMPA receptor (AMPAR) internalization provides a mechanism for long-term depression (LTD) in both hippocampal pyramidal neurons and cerebellar Purkinje cells (PCs). Cerebellar LTD at the parallel fiber (PF)-PC synapse is the underlying basis of motor learning and requires AMPAR activation
New On-line Mass Spectrometer for Identification of Reaction Products in the Aqueous Phase: Application to the OH-oxidation of N-methylpyrrolidone under Atmospheric Conditions.
Poulain L, et al.
Environmental Simulation Chambers: Application to Atmospheric Chemical Processes, 83-96 (2006)
Aurélie Leverrier et al.
Phytochemistry, 72(11-12), 1443-1452 (2011-05-10)
An in vitro biological screening of Malaysian plants allowed the selection of several species with a significant binding affinity for the antiapoptotic protein Bcl-xL. The chemical investigation of Endiandra kingiana led to the isolation of a series of polyketides named

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service