Skip to Content
Merck
All Photos(2)

Documents

147338

Sigma-Aldrich

Benzylidenemalononitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C(CN)2
CAS Number:
Molecular Weight:
154.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

83-85 °C (lit.)

SMILES string

N#C\C(=C/c1ccccc1)C#N

InChI

1S/C10H6N2/c11-7-10(8-12)6-9-4-2-1-3-5-9/h1-6H

InChI key

WAVNYPVYNSIHNC-UHFFFAOYSA-N

General description

Benzylidenemalononitrile undergoes rapid and catalyst-free solid/vapor reaction with primary alkyl amines to yield N-benzylidene-amine. It undergoes addition reaction with octyl- and decylzirconocene chloride. It is a potential substrate for soluble methane monooxygenase.

Application

Benzylidenemalononitrile was used in fluorescence-based assay for determination of soluble methane monooxygenase activity in solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanhong Liu et al.
The Journal of organic chemistry, 67(20), 7019-7028 (2002-10-02)
Direct addition of alkylzirconocenes to activated alkenes was found, for the first time. Octyl- and decylzirconocene chloride reacted with benzylidenemalononitrile to give the corresponding addition products after hydrolysis in 86% and 79% yield, respectively. Zirconacyclopentanes showed a similar reactivity toward
Liqi Shi et al.
Chemical communications (Cambridge, England), 50(7), 872-874 (2013-12-04)
A new rapid and catalyst-free solid/vapor reaction between benzylidenemalonate/benzylidenemalononitrile and primary alkyl amines was found. With these as sensory units of fluorescent polymers, probes for primary amine vapor with high sensitivity and selectivity were developed.
A R Miller et al.
Applied microbiology and biotechnology, 58(2), 183-188 (2002-03-06)
A fluorescence-based assay was developed to estimate soluble methane monooxygenase (sMMO) activity in solution. Whole cells of Methylosinus trichosporium OB3b expressing sMMO were used to oxidize various compounds to screen for fluorescent products. Of the 12 compounds tested, only coumarin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service