Skip to Content
Merck
All Photos(2)

Documents

N26806

Sigma-Aldrich

trans-β-Nitrostyrene

99%

Synonym(s):

trans-beta-Nitrostyrene, trans-1-Nitro-2-phenylethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHNO2
CAS Number:
Molecular Weight:
149.15
Beilstein:
1210066
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

250-260 °C (lit.)

mp

55-58 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)\C=C\c1ccccc1

InChI

1S/C8H7NO2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H/b7-6+

InChI key

PIAOLBVUVDXHHL-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellen Zandvoort et al.
Chembiochem : a European journal of chemical biology, 13(13), 1869-1873 (2012-08-02)
Serendipitous switch: While exploring cis-nitrostyrene as a potential electrophile in Michael-type addition reactions catalysed by the enzyme 4-oxalocrotonate tautomerase (4-OT), it was unexpectedly found that 4-OT catalyses the isomerisation of cis-nitrostyrene to trans-nitrostyrene (k(cat) /K(m) = 1.9×10(3)  M(-1)  s(-1) ).
Jens Martin Werner et al.
Journal of pharmacological and toxicological methods, 57(2), 131-137 (2007-12-19)
Induction of apoptosis is perceived as the main intention of drug regimens for tumour therapy. Thus, the concentration- and time-dependence of drug-induced apoptosis should be carefully evaluated for experimental as well as for standard anti-tumour agents. A main feature of
Junguk Park et al.
Biochemistry, 43(47), 15014-15021 (2004-11-24)
Protein tyrosine phosphatases (PTPs) catalyze the hydrolysis of phosphotyrosyl (pY) proteins to produce tyrosyl proteins and inorganic phosphate. Specific PTPs inhibitors provide useful tools for studying PTP function in signal transduction processes and potential treatment for human diseases such as
Hui Yang et al.
Organic & biomolecular chemistry, 10(16), 3229-3235 (2012-03-10)
(S)-Proline-catalyzed nitro-Michael additions of aldehydes and ketones to β-nitrostyrene were investigated computationally (MP2/6-311+G**//M06-2X/6-31G**). Contrary to what is usually assumed in organocatalysis, the lowest-energy transition states of proline-catalyzed nitro-Michael reactions do not necessarily involve the carboxylic acid group of the proline
Catalytic asymmetric intermolecular Stetter reactions of enolizable aldehydes with nitrostyrenes: computational study provides insight into the success of the catalyst.
Daniel A DiRocco et al.
Angewandte Chemie (International ed. in English), 51(10), 2391-2394 (2012-01-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service