Skip to Content
Merck

36928

1,2,3,5-Tetrachlorobenzene

PESTANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H2Cl4
CAS Number:
Molecular Weight:
215.89
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
211-217-7
Beilstein/REAXYS Number:
1618864
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1cc(Cl)c(Cl)c(Cl)c1

InChI

1S/C6H2Cl4/c7-3-1-4(8)6(10)5(9)2-3/h1-2H

InChI key

QZYNWJQFTJXIRN-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Peter Kämpfer et al.
International journal of systematic and evolutionary microbiology, 54(Pt 3), 749-751 (2004-05-15)
A Gram-positive, rod-shaped, non-spore-forming bacterium (B5(T)) was isolated from an enrichment culture that contained 1,2,3,5-tetrachlorobenzene as the sole source of carbon. On the basis of 16S rRNA gene sequence similarity studies, strain B5(T) was shown to belong to the family
B Z Fathepure et al.
Applied and environmental microbiology, 54(2), 327-330 (1988-02-01)
Hexachlorobenzene was dechlorinated to tri- and dichlorobenzenes in anaerobic sewage sludge. The complete biotransformation of 190 microM hexachlorobenzene (approximately 50 ppm) occurred within 3 weeks. The calculated rate of hexachlorobenzene dechlorination was 13.6 mumol liter-1 day-1. Hexachlorobenzene was dechlorinated via
M Parrini et al.
Bollettino della Societa italiana di biologia sperimentale, 66(7), 709-716 (1990-07-01)
The mutagenicity of halogenated benzenes, including three isomers of tri- and tetrachlorobenzenes (TCB, TeCB) was studied on male Swiss CD1 mice by MN test. The data presented show that all the halogenated benzenes tested were found to be clastogenic apart



Global Trade Item Number

SKUGTIN
36928-250MG04061835352883