Skip to Content
Merck
All Photos(1)

Key Documents

125377

Sigma-Aldrich

3-(Triethoxysilyl)propionitrile

97%

Synonym(s):

(2-Cyanoethyl)triethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3SiCH2CH2CN
CAS Number:
Molecular Weight:
217.34
Beilstein:
1776392
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

224 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](CCC#N)(OCC)OCC

InChI

1S/C9H19NO3Si/c1-4-11-14(12-5-2,13-6-3)9-7-8-10/h4-7,9H2,1-3H3

InChI key

GBQYMXVQHATSCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Triethoxysilyl)propionitrile (TESPN) is an organosilyl sol gel that can be used as a silane based coupling agent to improve the adhesion between metal-polymeric composite. It is non-toxic and cost efficient which makes it a suitable alternative to the existing chromate treatment of metals. It can be used in surface modification of silica based surfaces.

Application

3-(Triethoxysilyl)propionitrile (TESPN) may be coated on quartz and aluminum by the sol-gel process.
TESPN can functionalize silica nanoparticles for the fabrication of solid state electrolyte based dye sensitized solar cells (DSSCs) and for the development of PVDf based ultrafiltration membranes for antifouling applications. It can also be used in the synthesis of SBA-15 for the formation of mesoporous silica for drug delivery applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.9 °F - closed cup

Flash Point(C)

70.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silica nanoparticles as carriers of antifouling ligands for PVDF ultrafiltration membranes
Wu H, et al.
Journal of Membrane Science , 433(3), 135-151 (2013)
3-(Triethoxysilyl) propionitrile sol-gel coating
Li Y, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 61(7), 1675-1681 (2005)
3-(Triethoxysilyl)propionitrile sol?gel coating
Spectrochimica Acta Part A: Molecular Spectroscopy, 61(7), 1675-1681 (2005)
Quasi-solid state electrolytes with silica nanomaterial for high efficiency dye-sensitized solar cells
Jeon S, et al.
Rapid Communication in Photoscience, 2(3), 85-88 (2013)
Swelling-Agent-Free Synthesis of Siliceous and Functional Mesocellular Foam-Like Mesophases by Using a Carboxy-Terminated Triblock Copolymer
Hsu Y, et al.
Advances in Functional Materials, 18(12), 1799-1808 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service