Merck
  • The influence of pretreatment methods on saccharification of sugarcane bagasse by an enzyme extract from Chrysoporthe cubensis and commercial cocktails: A comparative study.
The influence of pretreatment methods on saccharification of sugarcane bagasse by an enzyme extract from Chrysoporthe cubensis and commercial cocktails: A comparative study.
Bioresource technology (2015-06-22)
Gabriela Piccolo Maitan-Alfenas, Evan Michael Visser, Rafael Ferreira Alfenas, Bráulio Ris G Nogueira, Guilherme Galvão de Campos, Adriane Ferreira Milagres, Ronald P de Vries, Valéria Monteze Guimarães
ABSTRACT
Biomass enzymatic hydrolysis depends on the pretreatment methods employed, the composition of initial feedstock and the enzyme cocktail used to release sugars for subsequent fermentation into ethanol. In this study, sugarcane bagasse was pretreated with 1% H2SO4 and 1% NaOH and the biomass saccharification was performed with 8% solids loading using 10 FPase units/g of bagasse of the enzymatic extract from Chrysoporthe cubensis and three commercial cocktails for a comparative study. Overall, the best glucose and xylose release was obtained from alkaline pretreated sugarcane bagasse. The C. cubensis extract promoted higher release of glucose (5.32 g/L) and xylose (9.00 g/L) than the commercial mixtures. Moreover, the C. cubensis extract presented high specific enzyme activities when compared to commercial cocktails mainly concerning to endoglucanase (331.84 U/mg of protein), β-glucosidase (29.48 U/mg of protein), β-xylosidase (2.95 U/mg of protein), pectinase (127.46 U/mg of protein) and laccase (2.49 U/mg of protein).
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Ammonium nitrate-14N2 solution, ~40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis