Merck
  • Mononuclear Polypyridylruthenium(II) Complexes with High Membrane Permeability in Gram-Negative Bacteria-in particular Pseudomonas aeruginosa.
Mononuclear Polypyridylruthenium(II) Complexes with High Membrane Permeability in Gram-Negative Bacteria-in particular Pseudomonas aeruginosa.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-06-05)
Anil K Gorle, Marshall Feterl, Jeffrey M Warner, Sebastian Primrose, Constantin C Constantinoiu, F Richard Keene, J Grant Collins
ABSTRACT
Ruthenium(II) complexes containing the tetradentate ligand bis[4(4'-methyl-2,2'-bipyridyl)]-1,n-alkane ("bbn "; n=10 and 12) have been synthesised and their geometric isomers separated. All [Ru(phen)(bbn )](2+) (phen=1,10-phenanthroline) complexes exhibited excellent activity against Gram-positive bacteria, but only the cis-α-[Ru(phen)(bb12 )](2+) species showed good activity against Gram-negative species. In particular, the cis-α-[Ru(phen)(bb12 )](2+) complex was two to four times more active than the cis-β-[Ru(phen)(bb12 )](2+) complex against the Gram-negative strains. The cis-α- and cis-β-[Ru(phen)(bb12 )](2+) complexes readily accumulated in the bacteria but, significantly, showed the highest level of uptake in Pseudomonas aeruginosa. Furthermore, the accumulation of the cis-α- and cis-β-[Ru(phen)(bb12 )](2+) complexes in P. aeruginosa was considerably greater than in Escherichia coli. The uptake of the cis-α-[Ru(phen)(bb12 )](2+) complex into live P. aeruginosa was confirmed by using fluorescence microscopy. The water/octanol partition coefficients (log P) were determined to gain understanding of the relative cellular uptake. The cis-α- and cis-β-[Ru(phen)(bbn )](2+) complexes exhibited relatively strong binding to DNA (Kb ≈10(6)  M(-1) ), but no significant difference between the geometric isomers was observed.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Potassium hexafluorophosphate, 99.5% trace metals basis
Sigma-Aldrich
Potassium hexafluorophosphate, ≥99%
Sigma-Aldrich
Ammonium hexafluorophosphate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium hexafluorophosphate, ≥95%
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D