Skip to Content
Merck
All Photos(1)

Key Documents

A1593

Sigma-Aldrich

Ampicillin

meets USP testing specifications

Synonym(s):

Ampicillin trihydrate, D-(−)-α-Aminobenzylpenicillin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O4S · 3H2O
CAS Number:
Molecular Weight:
403.45
Beilstein:
5399534
EC Number:
MDL number:
UNSPSC Code:
51281703
PubChem Substance ID:
NACRES:
NA.76

Agency

USP/NF
meets USP testing specifications

Quality Level

form

solid

color

white

mp

198-200 °C (dec.) (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

pharmaceutical (small molecule)

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccccc3)C(=O)N2[C@H]1C(O)=O

InChI

1S/C16H19N3O4S.3H2O/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;;;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);3*1H2/t9-,10-,11+,14-;;;/m1.../s1

InChI key

RXDALBZNGVATNY-CWLIKTDRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Used to select for ampicillin resistance in mutated and transformed cells.

Biochem/physiol Actions

A β-lactam antibiotic with an amino group side chain attached to the penicillin structure. Penicillin derivative that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Bactericidal only to growing Escherichia coli . Mode of resistance: Cleavage of β-lactam ring of ampicillin by β-lactamase. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.

Packaging

25g

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nai-Chi Chen et al.
PLoS pathogens, 11(10), e1005203-e1005203 (2015-10-23)
Betanodaviruses cause massive mortality in marine fish species with viral nervous necrosis. The structure of a T = 3 Grouper nervous necrosis virus-like particle (GNNV-LP) is determined by the ab initio method with non-crystallographic symmetry averaging at 3.6 Å resolution.
Kate S Baker et al.
The Lancet. Infectious diseases, 15(8), 913-921 (2015-05-06)
Shigellosis is an acute, severe bacterial colitis that, in high-income countries, is typically associated with travel to high-risk regions (Africa, Asia, and Latin America). Since the 1970s, shigellosis has also been reported as a sexually transmitted infection in men who
Jens T Van Praet et al.
The EMBO journal, 34(4), 466-474 (2015-01-21)
Antinuclear antibodies are a hallmark feature of generalized autoimmune diseases, including systemic lupus erythematosus and systemic sclerosis. However, the processes underlying the loss of tolerance against nuclear self-constituents remain largely unresolved. Using mice deficient in lymphotoxin and Hox11, we report
Ramendra K Singh et al.
European journal of medicinal chemistry, 45(3), 1078-1086 (2009-12-26)
Curcumin bioconjugates, viz. di-O-tryptophanylphenylalanine curcumin (2), di-O-decanoyl curcumin (3), di-O-pamitoyl curcumin (4), di-O-bis-(gamma,gamma)folyl curcumin (6), C(4)-ethyl-O-gamma-folyl curcumin (8) and 4-O-ethyl-O-gamma-folyl curcumin (10) have been synthesized and tested for their antibacterial and antiviral activities. The conjugates 2, 3, 4, 6 and
Marcel Weber et al.
PloS one, 9(6), e100000-e100000 (2014-06-21)
Several synthetic antibody phage display libraries have been created and used for the isolation of human monoclonal antibodies. The performance of antibody libraries, which is usually measured in terms of their ability to yield high-affinity binding specificities against target proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service