Skip to Content
Merck
All Photos(4)

Key Documents

201138

Sigma-Aldrich

Ammonium hexafluorophosphate

≥95%

Synonym(s):

Ammonium hexafluophosphate, Ammonium hexafluorophosphate V, Ammonium phosphorus hexafluoride, Ammoniumhexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4PF6
CAS Number:
Molecular Weight:
163.00
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38070413
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

form

solid

density

2.18 g/mL at 25 °C (lit.)

SMILES string

[H][N+]([H])([H])[H].F[P-](F)(F)(F)(F)F

InChI

1S/F6P.H3N/c1-7(2,3,4,5)6;/h;1H3/q-1;/p+1

InChI key

NIZXKAYXSNUDOU-UHFFFAOYSA-O

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium hexafluorophosphate (NH4PF6) can be used as a precursor for the preparation of:
  • Metal complexes, particularly iridium and rhodium complexes.       
  • Flame retardant cellulose-based materials.       
  • Cyclic amidinium and iminium ammonium hexafluorophosphate salts.        
  • The polymer-supported iridium isomerization catalyst for the selective trans isomerization of aryl allylic derivatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparations of iridium complexes containing phenanthroline ancillary ligands and electrical properties of cationic iridium-based light-emitting electrochemical cells
Park S, et al.
Surface and Interface Analysis : SIA, 44(11-12), 1479-1482 (2012)
A polymer-supported iridium catalyst for the stereoselective isomerisation of double bonds
Baxendale IR, et al.
Synlett, 2002(03), 0516-0518 (2002)
A simple and efficient one-step protocol for the preparation of alkyl-substituted ammonium tetrafluoroborate and hexafluorophosphate salts
Saba Shahrokh, et al.
Journal of Fluorine Chemistry, 153, 168-171 (2013)
Accelerated luminophore discovery through combinatorial synthesis.
Lowry, Michael S et al.
Journal of the American Chemical Society, 126(43), 14129-14135 (2004)
Homogeneous catalytic system for photoinduced hydrogen production utilizing iridium and rhodium complexes.
Cline, Eric D et al.
Inorganic Chemistry, 47(22), 10378-10388 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service