Merck
  • Bioorthogonal Labeling, Bioimaging, and Photocytotoxicity Studies of Phosphorescent Ruthenium(II) Polypyridine Dibenzocyclooctyne Complexes.
Bioorthogonal Labeling, Bioimaging, and Photocytotoxicity Studies of Phosphorescent Ruthenium(II) Polypyridine Dibenzocyclooctyne Complexes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-06-23)
Tommy Siu-Ming Tang, Alex Man-Hei Yip, Kenneth Yin Zhang, Hua-Wei Liu, Po Lam Wu, King Fai Li, Kok Wai Cheah, Kenneth Kam-Wing Lo
ABSTRACT
The synthesis, characterization, photophysics, lipophilicity, and cellular properties of new phosphorescent ruthenium(II) polypyridine complexes functionalized with a dibenzocyclooctyne (DIBO) or amine moiety [Ru(N^N)2 (L)](PF6 )2 are reported (L=4-(13-N-(3,4:7,8-dibenzocyclooctyne-5-oxycarbonyl) amino-4,7,10-trioxa-tridecanyl-aminocarbonyl-oxy-methyl)-4'-methyl-2,2'-bipyridine bpy-DIBO, N^N=2,2'-bipyridine bpy (1 a), 1,10-phenanthroline phen (2 a); L=4-(13-amino-4,7,10-trioxa-tridecanylaminocarbonyl-oxy-methyl)-4'-methyl-2,2'-bipyridine bpy-NH2 , N^N=bpy (1 b), phen (2 b)). The strain-promoted alkyne-azide cycloaddition (SPAAC) reaction of the DIBO complexes 1 a and 2 a with benzyl azide were studied. Also, the DIBO complexes 1 a and 2 a can selectively label N-azidoglycans located on the surface of CHO-K1 and A549 cells that were pretreated with 1,3,4,6-tetra-O-acetyl-N-azidoacetyl-D-mannosamine (Ac4 ManNAz). Additionally, the intracellular trafficking and localization of these biomolecules were monitored using laser-scanning confocal microscopy. Interestingly, the biolabeling and cellular uptake efficiency of the DIBO complexes 1 a and 2 a were cell-line dependent, as revealed by flow cytometry and ICP-MS. Furthermore, the complexes showed good biocompatibility toward the Ac4 ManNAz-pretreated cells in the dark, but exhibited photoinduced cytotoxicity due to the generation of singlet oxygen.
Sigma-Aldrich
Benzyl azide solution, ~0.5 M in dichloromethane, ≥95.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
4,4′-Dimethyl-2,2′-dipyridyl, 99.5%, purified by sublimation
Sigma-Aldrich
4,7,10-Trioxa-1,13-tridecanediamine, 97%
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, ≥99%
Sigma-Aldrich
4,4′-Dimethyl-2,2′-dipyridyl, 99%
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, 99%
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in THF
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in methylene chloride
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 1.0 M in THF
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%