Skip to Content
Merck
All Photos(2)

Key Documents

GF83525193

Ruthenium

Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%

Synonym(s):

Ruthenium, RU008010

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ru
CAS Number:
Molecular Weight:
101.07
MDL number:
UNSPSC Code:
12141739
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 835-251-93

resistivity

7.1 μΩ-cm, 0°C

L × W × thickness

25 mm × 2 mm × 2 mm

bp

3900 °C (lit.)

mp

2310 °C (lit.)

density

12.45 g/cm3 (lit.)

SMILES string

[Ru]

InChI

1S/Ru

InChI key

KJTLSVCANCCWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lutz Ackermann et al.
Topics in current chemistry, 292, 211-229 (2010-01-01)
Stoichiometric cycloruthenation reactions of substrates containing Lewis-basic functionalities set the stage for efficient ruthenium-catalyzed C-H bond functionalization reactions. Thereby, selective addition reactions of C-H bonds across alkenes or alkynes enabled atom-economical synthesis of substituted arenes. More recently, ruthenium-catalyzed direct arylation
Nicole L Fry et al.
Accounts of chemical research, 44(4), 289-298 (2011-03-03)
Nitric oxide (NO) can induce apoptosis (programmed cell death) at micromolar or higher doses. Although cell death via NO-induced apoptosis has been studied quite extensively, the targeted delivery of such doses of NO to infected or malignant tissues has not
Christian G Hartinger et al.
Current topics in medicinal chemistry, 11(21), 2688-2702 (2011-11-02)
Polynuclear compounds are a relatively new and successful approach in metal-based cancer chemotherapy as typified by the trinuclear Pt compound BBR3464 which was evaluated in clinical trials. In this review, we discuss newer developments of polynuclear ruthenium, osmium and gold
Martin R Gill et al.
Chemical Society reviews, 41(8), 3179-3192 (2012-02-09)
In the last few decades, coordination complexes based on d(6) metal centres and polypyridyl ligand architectures been developed as structure- and site-specific reversible DNA binding agents. Due to their attractive photophysical properties, much of this research has focused on complexes
A Bergamo et al.
Journal of inorganic biochemistry, 106(1), 90-99 (2011-11-25)
The study of metal complexes for the treatment of cancer diseases has resulted in the identification of some unique properties of ruthenium-based compounds. Among these inorganic-based agents, two of them, namely the ruthenium(III) drugs NAMI-A and KP1019 have undertaken with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service