Skip to Content
Merck
All Photos(3)

Key Documents

205249

Sigma-Aldrich

Di-tert-butyl dicarbonate

99%, for peptide synthesis, ReagentPlus®

Synonym(s):

Boc anhydride, Di-tert-butyl pyrocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3COCO]2O
CAS Number:
Molecular Weight:
218.25
Beilstein:
1911173
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Product Name

Di-tert-butyl dicarbonate, ReagentPlus®, 99%

Quality Level

product line

ReagentPlus®

Assay

99%

form

solid or liquid

refractive index

n20/D 1.409 (lit.)

bp

56-57 °C/0.5 mmHg (lit.)

mp

23 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

functional group

carbonate

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

InChI

1S/C10H18O5/c1-9(2,3)14-7(11)13-8(12)15-10(4,5)6/h1-6H3

InChI key

DYHSDKLCOJIUFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aminomethyl allene was prepared by reaction of Boc propargyl amine with formaldehyde, diisopropylamine and copper bromide as part of a series of Bovine Plasma Amine Oxidase inactivators.
Protection of alcohols as Boc derivatives via Lewis acid catalysis.
Reagent for the introduction of the Boc protecting group.
Reagent for the preparation of Boc protected amines.
Tris-Boc protected hydrazine allows preparation of Fmoc ester in a chromophoric reagent to monitor solid-phase aldehydes.

Automated Boc protection and deprotection can be done using Synple Automated Synthesis Platform (SYNPLE-SC002), Boc protection cartridges ((SYNPLE-B001), (SYNPLE-B002), and Boc deprotection cartridges (SYNPLE-B011)

Warning

Hydrolyzes to t-butanol and CO2; causes internal pressure in bottle if exposed to moisture.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon K Shannon et al.
The Journal of organic chemistry, 69(14), 4586-4594 (2004-07-03)
A direct method for quantifying solid-phase aldehydes has been developed, using a new reagent, 4-(9-fluorenylmethyloxycarbonyl)phenylhydrazine (FmPH). The FmPH reagent was synthesized in three steps (24% overall yield) from commercially available p-hydrazinobenzoic acid. Resin-bound aldehydes reacted quantitatively with FmPH, in the
Synthetic Communications, 23, 1443-1443 (1993)
Chunhua Qiao et al.
Journal of the American Chemical Society, 126(25), 8038-8045 (2004-06-24)
Propargylic and activated allylic amines are known to inactivate the quinone-dependent plasma amine oxidases, possibly through active-site modification by the alpha,beta-unsaturated aldehyde turnover products. Although homopropargylamine (1-amino-3-butyne, 1) is a nonobvious candidate as a mechanism-based inhibitor, 1 was found to
Synlett, 2104-2104 (2006)
Jason C Green et al.
Organic letters, 13(20), 5500-5503 (2011-09-21)
Total syntheses of two structures purported as (+)-heliananes were completed in six pots. Spectral comparisons, between the synthetic and natural compounds, revealed a misassignment of the eight-membered ring in the heliananes. The key step in the syntheses of the proposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service