Merck
  • Detection of volatile organic compounds indicative of human presence in the air.
Detection of volatile organic compounds indicative of human presence in the air.
Journal of separation science (2015-05-07)
Jae Kwak, Brian A Geier, Maomian Fan, Sanjay A Gogate, Sage A Rinehardt, Brandy S Watts, Claude C Grigsby, Darrin K Ott
ABSTRACT
Volatile organic compounds were collected and analyzed from a variety of indoor and outdoor air samples to test whether human-derived compounds can be readily detected in the air and if they can be associated with human occupancy or presence. Compounds were captured with thermal desorption tubes and then analyzed by gas chromatography with mass spectrometry. Isoprene, a major volatile organic compound in exhaled breath, was shown to be the best indicator of human presence. Acetone, another major breath-borne compound, was higher in unoccupied or minimally occupied areas than in human-occupied areas, indicating that its majority may be derived from exogenous sources. The association of endogenous skin-derived compounds with human occupancy was not significant. In contrast, numerous compounds that are found in foods and consumer products were detected at elevated levels in the occupied areas. Our results revealed that isoprene and many exogenous volatile organic compounds consumed by humans are emitted at levels sufficient for detection in the air, which may be indicative of human presence.
Sigma-Aldrich
1-Bromo-4-fluorobenzene, 99%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Phenylethanol, ≥99.0% (GC)
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Chlorobenzene-d5, 99 atom % D
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Phenethyl alcohol, natural, ≥99%, FCC, FG