Skip to Content
Merck
All Photos(1)

Key Documents

733458

Sigma-Aldrich

2-Propanol

99.999% trace metals basis, electronic grade

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23
grade:
electronic grade
Assay:
99.999% trace metals basis
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Product Name

2-Propanol, electronic grade, 99.999% trace metals basis

grade

electronic grade

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

99.999% trace metals basis

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

impurities

≤0.05% water
water

color

colorless

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

water: soluble (completely)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Propanol (IPA) is an electronic graded solvent that is majorly used in the rinsing of semi-conductors, metals and organic polymers.

Application

IPA can be used to rinse a variety of materials like gold-titanium probes, methanol fuel cells, organic semiconductors and silicon wafers.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sol-gel synthesis and characterization of nanocrystalline ZnO powders.
Ristic M, et al.
Journal of alloys and compounds, 397(1-2), L1-L4 (2005)
Oriented UiO-66 thin films through solution shearing.
Ghorbanpour A, et al.
CrystEngComm, 20(3), 294-300 (2018)
Effects of ethanol in methanol fuel on the performance of membrane electrode assemblies for direct methanol fuel cells.
Yang S, et al.
Journal of Industrial and Engineering Chemistry (2018)
Fabrication of electrically contacted plasmonic Schottky nanoantennas on silicon.
Alavirad M, et al.
Chinese Optics Letters, 16(5), 050007-050007 (2018)
Tattoo-Paper Transfer as a Versatile Platform for All-Printed Organic Edible Electronics.
Bonacchini GE, et al.
Advanced Materials, 30(14), 1706091-1706091 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service