Skip to Content
Merck
All Photos(3)

Key Documents

278475

Sigma-Aldrich

2-Propanol

99.5%, anhydrous

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)
Pricing and availability is not currently available.

Product Name

2-Propanol, anhydrous, 99.5%

grade

anhydrous

Quality Level

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

99.5%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

color

colorless

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

water: soluble (completely)

density

0.785 g/mL at 25 °C (lit.)

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Propanol may be used as a solvent in the preparation of cyclic ureas by reacting carbon dioxide with diamines in the presence of pure cerium oxide (CeO2).[1] It may also be used as an alternate precursor to acetone for synthesizing methyl isobutyl ketone.[2]

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Pure-Pac® II containers require the Micromatic MicroValve coupler for dispensing solvents, Z560723.

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step MIBK synthesis: a new process from 2-propanol
Di Cosimo JI, et al
J. Catal., 208(1), 114-123 (2002)
Fabrication and characterization of electrospun titania nanofibers
Chandrasekar R, et al.
J. Mater. Sci., 44(5), 1198-1205 (2009)
Qing-Fei Yin et al.
Methods in molecular biology (Clifton, N.J.), 1206, 69-80 (2014-09-23)
Most of mRNAs and well-characterized long noncoding RNAs are shaped with 5' cap and 3' poly(A) tail. Thereby, conventional transcriptome analysis typically involved the enrichment of poly(A)+ RNAs by oligo(dT) selection. However, accumulated lines of evidence suggest that there are
Highly efficient synthesis of cyclic ureas from CO2 and diamines by a pure CeO2 catalyst using a 2-propanol solvent.
Tamura M, et al.
Green Chemistry, 15(6), 1567-1577 (2013)
One-step MIBK synthesis: a new process from 2-propanol.
Di Cosimo JI, et al.
J. Catal., 208(1), 114-123 (2002)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Hi, I am looking for anhydrous 2-propanol to use in glovebox . Could you please recommend whether this should be fine?

    1 answer
    1. This product is packaged under nitrogen and is in Sure/Seal to ensure the water content remains very low. This material can be brought into a glove box with the proper care. It is essential to follow appropriate dispensing procedures when handling air-sensitive products. To assist you in this, please consult “Technical Bulletin AL-134: Handling Air-Sensitive Reagents.

      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/685/583/al_techbull_al134.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service