Merck
  • Products and Mechanism of the Reaction of 1-Pentadecene with NO3 Radicals and the Effect of a -ONO2 Group on Alkoxy Radical Decomposition.
Products and Mechanism of the Reaction of 1-Pentadecene with NO3 Radicals and the Effect of a -ONO2 Group on Alkoxy Radical Decomposition.
The journal of physical chemistry. A (2015-10-06)
Geoffrey K Yeh, Megan S Claflin, Paul J Ziemann
ABSTRACT
The linear C15 alkene, 1-pentadecene, was reacted with NO3 radicals in a Teflon environmental chamber and yields of secondary organic aerosol (SOA) and particulate β-hydroxynitrates, β-carbonylnitrates, and organic peroxides (β-nitrooxyhydroperoxides + dinitrooxyperoxides) were quantified using a variety of methods. Reaction occurs almost solely by addition of NO3 to the C═C double bond and measured yields of β-hydroxynitrate isomers indicate that 92% of addition occurs at the terminal carbon. Molar yields of reaction products determined from measurements, a proposed reaction mechanism, and mass-balance considerations were 0.065 for β-hydroxynitrates (0.060 and 0.005 for 1-nitrooxy-2-hydroxypentadecane and 1-hydroxy-2-nitrooxypentadecane isomers), 0.102 for β-carbonylnitrates, 0.017 for organic peroxides, 0.232 for β-nitrooxyalkoxy radical isomerization products, and 0.584 for tetradecanal and formaldehyde, the volatile C14 and C1 products of β-nitrooxyalkoxy radical decomposition. Branching ratios for decomposition and isomerization of β-nitrooxyalkoxy radicals were 0.716 and 0.284 and should be similar for other linear 1-alkenes ≥ C6 whose alkyl chains are long enough to allow for isomerization to occur. These branching ratios have not been measured previously, and they differ significantly from those estimated using structure-activity relationships, which predict >99% isomerization. It appears that the presence of a -ONO2 group adjacent to an alkoxy radical site greatly enhances the rate of decomposition relative to isomerization, which is otherwise negligible, and that the effect is similar to that of a -OH group. The results provide insight into the effects of molecular structure on mechanisms of oxidation of volatile organic compounds and should be useful for improving structure-activity relationships that are widely used to predict the fate of these compounds in the atmosphere and for modeling SOA formation and aging.
Sigma-Aldrich
Luperox® A75FP, Benzoyl peroxide, 75% remainder water, contains 25 wt. % water as stabilizer, 75%
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
2-Ethylhexyl nitrate, 97%
Sigma-Aldrich
Benzoyl peroxide blend with dicyclohexyl phthalate, suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.
Sigma-Aldrich
1-Pentadecene, 98%
Sigma-Aldrich
Dioctyl sebacate, AldrichCPR
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2,3-Dimethyl-2-butene, 98%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
2,3-Dimethyl-2-butene, ≥99%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%