Skip to Content
Merck

Skip To

L092003

Ethyl acetate

Synonym(s):

EtOAc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
EC Number:
205-500-4
UNSPSC Code:
12352108
MDL number:
Beilstein/REAXYS Number:
506104
NACRES:
NA.01
Bp:
76.5-77.5 °C (lit.)
Vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

water: soluble

density

0.902 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.008631.096231.10972
solubility

water: soluble

solubility

85.3 g/L

solubility

85.3 g/L

solubility

85.3 g/L

form

liquid

form

liquid

form

liquid

form

liquid

refractive index

n20/D 1.3720 (lit.)

refractive index

n20/D 1.3720 (lit.)

refractive index

n20/D 1.3720 (lit.)

refractive index

n20/D 1.3720 (lit.)

mp

−84 °C (lit.)

mp

−84 °C (lit.)

mp

−84 °C (lit.)

mp

−84 °C (lit.)

vapor pressure

73 mmHg ( 20 °C)

vapor pressure

73 mmHg ( 20 °C)

vapor pressure

73 mmHg ( 20 °C)

vapor pressure

73 mmHg ( 20 °C)

autoignition temp.

801 °F

autoignition temp.

801 °F

autoignition temp.

801 °F

autoignition temp.

801 °F


Still not finding the right product?

Explore all of our products under Ethyl acetate


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters